Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46857482-46858079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
UniGene
Hs.517939
Gene Chromosome Location
Chr.3: 46857867-46863483 on Build GRCh38
Transcripts
NM_000258.2

SNP Detail

SNP ID Chromosome Location
rs889802255 Chr.3: 46857762-46857762 on Build GRCh38
rs757528575 Chr.3: 46857989-46857989 on Build GRCh38
rs1027663046 Chr.3: 46857648-46857648 on Build GRCh38
rs886058581 Chr.3: 46857910-46857910 on Build GRCh38
rs570761980 Chr.3: 46857521-46857521 on Build GRCh38
rs773170066 Chr.3: 46857808-46857810 on Build GRCh38
rs1021372893 Chr.3: 46857941-46857941 on Build GRCh38
rs967106687 Chr.3: 46857952-46857952 on Build GRCh38
rs1028308912 Chr.3: 46857550-46857552 on Build GRCh38
rs984886455 Chr.3: 46857677-46857677 on Build GRCh38
rs933024341 Chr.3: 46857990-46857990 on Build GRCh38
rs528503721 Chr.3: 46857755-46857755 on Build GRCh38
rs993542091 Chr.3: 46857748-46857748 on Build GRCh38
rs78422374 Chr.3: 46857745-46857745 on Build GRCh38
rs578068639 Chr.3: 46857646-46857646 on Build GRCh38
rs1050738387 Chr.3: 46857623-46857623 on Build GRCh38
rs994773872 Chr.3: 46857836-46857836 on Build GRCh38
rs764865767 Chr.3: 46857975-46857975 on Build GRCh38
rs953115947 Chr.3: 46857942-46857942 on Build GRCh38
rs1034295388 Chr.3: 46857886-46857886 on Build GRCh38
rs1042973 Chr.3: 46858026-46858026 on Build GRCh38
rs556971059 Chr.3: 46857966-46857966 on Build GRCh38
rs893028278 Chr.3: 46857517-46857517 on Build GRCh38
rs969813921 Chr.3: 46857691-46857691 on Build GRCh38
rs1049793812 Chr.3: 46857760-46857760 on Build GRCh38
rs897216643 Chr.3: 46857818-46857818 on Build GRCh38
rs952979370 Chr.3: 46857661-46857662 on Build GRCh38
rs756317670 Chr.3: 46857939-46857939 on Build GRCh38
rs1019679019 Chr.3: 46857582-46857582 on Build GRCh38
rs1012491889 Chr.3: 46857957-46857957 on Build GRCh38
rs932973174 Chr.3: 46857619-46857619 on Build GRCh38
rs946706373 Chr.3: 46857682-46857698 on Build GRCh38
rs748065479 Chr.3: 46857600-46857603 on Build GRCh38
rs534585583 Chr.3: 46857527-46857527 on Build GRCh38
rs574063433 Chr.3: 46857790-46857790 on Build GRCh38
rs543352649 Chr.3: 46857658-46857658 on Build GRCh38
rs1027135879 Chr.3: 46857937-46857937 on Build GRCh38
rs868516602 Chr.3: 46857657-46857657 on Build GRCh38
rs958859807 Chr.3: 46857894-46857894 on Build GRCh38
rs1037845792 Chr.3: 46857715-46857715 on Build GRCh38
rs1008310328 Chr.3: 46857555-46857555 on Build GRCh38
rs759915302 Chr.3: 46857699-46857699 on Build GRCh38
rs989958816 Chr.3: 46857917-46857917 on Build GRCh38
rs1002615006 Chr.3: 46857857-46857857 on Build GRCh38
rs546418537 Chr.3: 46858042-46858042 on Build GRCh38
rs1008192587 Chr.3: 46857774-46857774 on Build GRCh38
rs13081285 Chr.3: 46857803-46857803 on Build GRCh38
rs922925834 Chr.3: 46857603-46857603 on Build GRCh38
rs889915066 Chr.3: 46857551-46857551 on Build GRCh38
rs768068456 Chr.3: 46857721-46857721 on Build GRCh38
rs756462246 Chr.3: 46857955-46857955 on Build GRCh38
rs754150859 Chr.3: 46857977-46857977 on Build GRCh38
rs546968708 Chr.3: 46858003-46858003 on Build GRCh38
rs942237459 Chr.3: 46858038-46858038 on Build GRCh38
rs1052927456 Chr.3: 46857743-46857745 on Build GRCh38
rs541549915 Chr.3: 46857797-46857797 on Build GRCh38

View Full Product Details