Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V0A2
Entrez Gene ID
23545
Gene Name
ATPase H+ transporting V0 subunit a2
Gene Aliases
A2, ARCL, ARCL2A, ATP6A2, ATP6N1D, J6B7, RTF, STV1, TJ6, TJ6M, TJ6S, VPH1, WSS
Chromosome Location
Chr.12: 123712318 - 123761755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012463.3 NP_036595.2 13 - 14 1850 61
GenBank mRNA AF112972.1 - 13 - 14 1607 61
AK289391.1 - 13 - 14 1765 61
BC068531.1 - 13 - 14 1850 61

View Full Product Details