The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136115.2 | NP_001129587.1 | 1 - 2 | 315 | 73 |
NM_015176.3 | NP_055991.1 | 1 - 2 | 315 | 73 | |
NR_049764.1 | 1 - 2 | 315 | 73 | ||
GenBank mRNA | AB007952.1 | - | 1 - 2 | 66 | 73 |
AK001628.1 | - | 1 - 2 | 291 | 73 | |
AK303381.1 | - | 1 - 2 | 283 | 73 | |
AK307892.1 | - | 1 - 2 | 287 | 73 | |
BC031576.1 | - | 1 - 2 | 284 | 73 | |
BP249147.1 | - | 1 - 2 | 313 | 73 | |
DB136175.1 | - | 1 - 2 | 316 | 73 |