Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
Chromosome Location
Chr.5: 96759245 - 96935983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040458.1 NP_001035548.1 2 - 3 868 69
NM_001198541.1 NP_001185470.1 2 - 3 626 69
NM_016442.3 NP_057526.3 2 - 3 868 69
XM_005272015.4 XP_005272072.1 2 - 3 819 69
XM_005272016.4 XP_005272073.1 2 - 3 779 69
XM_011543480.2 XP_011541782.1 6 - 7 3088 69
XM_011543481.2 XP_011541783.1 6 - 7 3085 69
XM_011543484.2 XP_011541786.1 6 - 7 3084 69
XM_011543485.2 XP_011541787.1 5 - 6 2904 69
XM_011543486.2 XP_011541788.1 6 - 7 3088 69
XM_017009580.1 XP_016865069.1 7 - 8 3144 69
XM_017009581.1 XP_016865070.1 5 - 6 2930 69
XM_017009582.1 XP_016865071.1 2 - 3 603 69
XM_017009583.1 XP_016865072.1 2 - 3 814 69
GenBank mRNA AB011097.2 - 1 - 2 542 69
AF106037.1 - 2 - 3 583 69
AF183569.1 - 2 - 3 588 69
AF222340.1 - 2 - 3 608 69
AK291636.1 - 2 - 3 618 69
AY028806.1 - 2 - 3 618 69
AY028807.1 - 2 - 3 618 69
AY358590.1 - 2 - 3 606 69
BC030775.1 - 2 - 3 620 69
JX962699.1 - 1 - 2 521 69
JX962700.1 - 1 - 2 521 69
JX962701.1 - 1 - 2 521 69
JX962702.1 - 1 - 2 521 69
KM357870.1 - 1 - 2 521 69
KM357871.1 - 1 - 2 521 69
KM357872.1 - 1 - 2 521 69
KM357873.1 - 1 - 2 521 69
KM357874.1 - 1 - 2 521 69
KM357875.1 - 1 - 2 521 69
KM357876.1 - 1 - 2 521 69
KM357877.1 - 1 - 2 521 69
KM357878.1 - 1 - 2 521 69
KM357879.1 - 1 - 2 521 69
KM357880.1 - 1 - 2 521 69
KM357881.1 - 1 - 2 521 69
KM357882.1 - 1 - 2 521 69
KM357883.1 - 1 - 2 521 69
KM357884.1 - 1 - 2 521 69
KM357885.1 - 1 - 2 521 69
KM357886.1 - 1 - 2 521 69
KM357887.1 - 1 - 2 521 69
KM357888.1 - 1 - 2 521 69
KM357889.1 - 1 - 2 521 69
KM357890.1 - 1 - 2 521 69
KM357891.1 - 1 - 2 521 69

View Full Product Details