The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297755.1 | NP_001284684.1 | 4 - 5 | 1447 | 115 |
NM_001297756.1 | NP_001284685.1 | 5 - 6 | 1648 | 115 | |
NM_133636.3 | NP_598375.2 | 5 - 6 | 1648 | 115 | |
XM_005262711.1 | XP_005262768.1 | 5 - 6 | 1509 | 115 | |
XM_005262713.2 | XP_005262770.1 | 5 - 6 | 1738 | 115 | |
XM_006714076.2 | XP_006714139.1 | 5 - 6 | 1610 | 115 | |
XM_011531580.2 | XP_011529882.1 | 5 - 6 | 1610 | 115 | |
XM_017007679.1 | XP_016863168.1 | 4 - 5 | 1308 | 115 | |
XM_017007680.1 | XP_016863169.1 | 4 - 5 | 1537 | 115 | |
XM_017007681.1 | XP_016863170.1 | 5 - 6 | 1610 | 115 | |
XM_017007682.1 | XP_016863171.1 | 5 - 6 | 1716 | 115 | |
XM_017007683.1 | XP_016863172.1 | 3 - 4 | 835 | 115 | |
GenBank mRNA | AF436845.1 | - | 5 - 6 | 1609 | 115 |
AK308395.1 | - | 4 - 5 | 1387 | 115 | |
AK312725.1 | - | 5 - 6 | 1612 | 115 | |
AL512702.1 | - | 3 - 4 | 299 | 115 | |
CK000270.1 | - | 2 - 3 | 248 | 115 |