Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35575339-35575917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378137 c.1029T>G p.G343G Chr.17: 35575833-35575833 on Build GRCh38
7163081 c.1037C>A p.T346K Chr.17: 35575825-35575825 on Build GRCh38
6744099 c.979C>T p.R327C Chr.17: 35575883-35575883 on Build GRCh38
6744098 c.1032T>C p.Y344Y Chr.17: 35575830-35575830 on Build GRCh38
977984 c.1051C>A p.L351M Chr.17: 35575811-35575811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751033919 Chr.17: 35575743-35575743 on Build GRCh38
rs761445143 Chr.17: 35575890-35575890 on Build GRCh38
rs113891712 Chr.17: 35575485-35575485 on Build GRCh38
rs756694664 Chr.17: 35575744-35575744 on Build GRCh38
rs763619631 Chr.17: 35575738-35575738 on Build GRCh38
rs912131686 Chr.17: 35575653-35575653 on Build GRCh38
rs748308886 Chr.17: 35575811-35575811 on Build GRCh38
rs989722936 Chr.17: 35575466-35575466 on Build GRCh38
rs572785120 Chr.17: 35575727-35575727 on Build GRCh38
rs780946763 Chr.17: 35575747-35575747 on Build GRCh38
rs762716335 Chr.17: 35575402-35575402 on Build GRCh38
rs756569847 Chr.17: 35575765-35575765 on Build GRCh38
rs751429784 Chr.17: 35575883-35575883 on Build GRCh38
rs555645908 Chr.17: 35575684-35575684 on Build GRCh38
rs760739894 Chr.17: 35575853-35575853 on Build GRCh38
rs895745225 Chr.17: 35575525-35575525 on Build GRCh38
rs758276967 Chr.17: 35575733-35575733 on Build GRCh38
rs941358133 Chr.17: 35575884-35575884 on Build GRCh38
rs966726269 Chr.17: 35575726-35575726 on Build GRCh38
rs570189667 Chr.17: 35575554-35575554 on Build GRCh38
rs1037590 Chr.17: 35575616-35575616 on Build GRCh38
rs928085463 Chr.17: 35575802-35575802 on Build GRCh38
rs570468178 Chr.17: 35575425-35575425 on Build GRCh38
rs762912502 Chr.17: 35575865-35575865 on Build GRCh38
rs905163839 Chr.17: 35575484-35575484 on Build GRCh38
rs899106737 Chr.17: 35575663-35575663 on Build GRCh38
rs200789791 Chr.17: 35575815-35575815 on Build GRCh38
rs772028045 Chr.17: 35575841-35575841 on Build GRCh38
rs866743376 Chr.17: 35575549-35575549 on Build GRCh38
rs754471266 Chr.17: 35575585-35575585 on Build GRCh38
rs886052830 Chr.17: 35575447-35575447 on Build GRCh38
rs752689493 Chr.17: 35575746-35575746 on Build GRCh38
rs1055712098 Chr.17: 35575517-35575517 on Build GRCh38
rs915485976 Chr.17: 35575754-35575754 on Build GRCh38
rs988425243 Chr.17: 35575729-35575729 on Build GRCh38
rs1045289989 Chr.17: 35575487-35575487 on Build GRCh38
rs1047155507 Chr.17: 35575848-35575848 on Build GRCh38
rs539591621 Chr.17: 35575445-35575445 on Build GRCh38
rs780501878 Chr.17: 35575782-35575782 on Build GRCh38
rs532390390 Chr.17: 35575681-35575681 on Build GRCh38
rs555997949 Chr.17: 35575494-35575494 on Build GRCh38
rs954551964 Chr.17: 35575735-35575735 on Build GRCh38
rs961888073 Chr.17: 35575569-35575569 on Build GRCh38
rs986241167 Chr.17: 35575629-35575629 on Build GRCh38
rs752715446 Chr.17: 35575665-35575665 on Build GRCh38
rs912165834 Chr.17: 35575385-35575385 on Build GRCh38
rs987184149 Chr.17: 35575426-35575426 on Build GRCh38
rs573245836 Chr.17: 35575427-35575427 on Build GRCh38
rs557834416 Chr.17: 35575882-35575882 on Build GRCh38
rs981025397 Chr.17: 35575800-35575800 on Build GRCh38
rs769244620 Chr.17: 35575786-35575786 on Build GRCh38
rs1005986145 Chr.17: 35575608-35575608 on Build GRCh38
rs959892912 Chr.17: 35575362-35575362 on Build GRCh38
rs200283718 Chr.17: 35575860-35575860 on Build GRCh38
rs996017117 Chr.17: 35575673-35575673 on Build GRCh38
rs764657253 Chr.17: 35575874-35575875 on Build GRCh38
rs779530147 Chr.17: 35575794-35575794 on Build GRCh38
rs939380551 Chr.17: 35575562-35575562 on Build GRCh38
rs915501223 Chr.17: 35575467-35575467 on Build GRCh38
rs749833303 Chr.17: 35575784-35575784 on Build GRCh38
rs267608184 Chr.17: 35575813-35575815 on Build GRCh38
rs887088674 Chr.17: 35575593-35575593 on Build GRCh38

View Full Product Details