Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9629828-9630405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024027 c.252G>T p.E84D Chr.2: 9630201-9630201 on Build GRCh38
6824612 c.191G>T p.S64I Chr.2: 9630262-9630262 on Build GRCh38
4464962 c.135C>T p.S45S Chr.2: 9630318-9630318 on Build GRCh38
6824608 c.206C>A p.T69N Chr.2: 9630247-9630247 on Build GRCh38
6584199 c.270G>A p.L90L Chr.2: 9630183-9630183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960357310 Chr.2: 9630272-9630272 on Build GRCh38
rs372377118 Chr.2: 9630130-9630130 on Build GRCh38
rs1055714443 Chr.2: 9630007-9630007 on Build GRCh38
rs920125450 Chr.2: 9629982-9629982 on Build GRCh38
rs749499495 Chr.2: 9630242-9630242 on Build GRCh38
rs746966183 Chr.2: 9630263-9630263 on Build GRCh38
rs986378939 Chr.2: 9629908-9629908 on Build GRCh38
rs756561669 Chr.2: 9630340-9630340 on Build GRCh38
rs13417081 Chr.2: 9629921-9629921 on Build GRCh38
rs1027369792 Chr.2: 9630210-9630210 on Build GRCh38
rs755484324 Chr.2: 9630213-9630213 on Build GRCh38
rs11544465 Chr.2: 9630236-9630236 on Build GRCh38
rs938210264 Chr.2: 9630006-9630006 on Build GRCh38
rs890405510 Chr.2: 9630089-9630089 on Build GRCh38
rs777649657 Chr.2: 9630381-9630381 on Build GRCh38
rs894533232 Chr.2: 9629871-9629871 on Build GRCh38
rs748029699 Chr.2: 9630369-9630369 on Build GRCh38
rs769631927 Chr.2: 9630373-9630373 on Build GRCh38
rs966744955 Chr.2: 9630106-9630106 on Build GRCh38
rs1013398204 Chr.2: 9629877-9629877 on Build GRCh38
rs1043086029 Chr.2: 9630070-9630070 on Build GRCh38
rs762606877 Chr.2: 9630177-9630177 on Build GRCh38
rs550294970 Chr.2: 9629931-9629931 on Build GRCh38
rs976217680 Chr.2: 9629935-9629935 on Build GRCh38
rs952825272 Chr.2: 9629991-9629991 on Build GRCh38
rs748020959 Chr.2: 9630312-9630312 on Build GRCh38
rs750464603 Chr.2: 9630356-9630356 on Build GRCh38
rs566200608 Chr.2: 9630040-9630040 on Build GRCh38
rs555195769 Chr.2: 9630047-9630047 on Build GRCh38
rs147641429 Chr.2: 9629993-9629993 on Build GRCh38
rs368778993 Chr.2: 9630108-9630108 on Build GRCh38
rs1027561217 Chr.2: 9629903-9629903 on Build GRCh38
rs778901859 Chr.2: 9630253-9630253 on Build GRCh38
rs896627860 Chr.2: 9630021-9630021 on Build GRCh38
rs934469948 Chr.2: 9630079-9630079 on Build GRCh38
rs1020766289 Chr.2: 9630382-9630382 on Build GRCh38
rs768534041 Chr.2: 9630267-9630267 on Build GRCh38
rs377250330 Chr.2: 9630342-9630342 on Build GRCh38
rs778991719 Chr.2: 9630355-9630355 on Build GRCh38
rs1017557119 Chr.2: 9630354-9630354 on Build GRCh38
rs777744668 Chr.2: 9630127-9630127 on Build GRCh38
rs372978538 Chr.2: 9630336-9630336 on Build GRCh38
rs567962081 Chr.2: 9629904-9629904 on Build GRCh38
rs746945960 Chr.2: 9630140-9630140 on Build GRCh38
rs776325430 Chr.2: 9630281-9630281 on Build GRCh38
rs762624973 Chr.2: 9630318-9630318 on Build GRCh38
rs983037522 Chr.2: 9629999-9629999 on Build GRCh38
rs141887759 Chr.2: 9630368-9630368 on Build GRCh38
rs756308327 Chr.2: 9630119-9630119 on Build GRCh38
rs908772629 Chr.2: 9630008-9630008 on Build GRCh38
rs1052978395 Chr.2: 9630088-9630088 on Build GRCh38
rs1037601336 Chr.2: 9630178-9630178 on Build GRCh38
rs982921555 Chr.2: 9630257-9630257 on Build GRCh38
rs575123175 Chr.2: 9630096-9630096 on Build GRCh38
rs769460753 Chr.2: 9630315-9630315 on Build GRCh38
rs948863925 Chr.2: 9630061-9630061 on Build GRCh38
rs201957113 Chr.2: 9630330-9630330 on Build GRCh38
rs539034877 Chr.2: 9629966-9629966 on Build GRCh38
rs202138500 Chr.2: 9630234-9630234 on Build GRCh38
rs757416206 Chr.2: 9630353-9630353 on Build GRCh38
rs773949858 Chr.2: 9630326-9630326 on Build GRCh38
rs111612155 Chr.2: 9629864-9629864 on Build GRCh38
rs748332272 Chr.2: 9630116-9630116 on Build GRCh38
rs998733894 Chr.2: 9629920-9629920 on Build GRCh38
rs752112912 Chr.2: 9630208-9630208 on Build GRCh38
rs776433787 Chr.2: 9630165-9630165 on Build GRCh38
rs369852702 Chr.2: 9630176-9630176 on Build GRCh38
rs781724742 Chr.2: 9630110-9630122 on Build GRCh38
rs756400871 Chr.2: 9630232-9630232 on Build GRCh38
rs764907087 Chr.2: 9630174-9630174 on Build GRCh38
rs755457718 Chr.2: 9630109-9630109 on Build GRCh38
rs371355879 Chr.2: 9630152-9630152 on Build GRCh38
rs528396693 Chr.2: 9629850-9629850 on Build GRCh38
rs1008533212 Chr.2: 9629937-9629937 on Build GRCh38
rs774239040 Chr.2: 9630199-9630199 on Build GRCh38
rs770978736 Chr.2: 9630136-9630136 on Build GRCh38
rs754065297 Chr.2: 9630351-9630351 on Build GRCh38
rs199958721 Chr.2: 9630214-9630214 on Build GRCh38
rs375839408 Chr.2: 9630133-9630133 on Build GRCh38
rs753064140 Chr.2: 9630225-9630225 on Build GRCh38
rs539717165 Chr.2: 9630300-9630300 on Build GRCh38
rs1017922046 Chr.2: 9630093-9630093 on Build GRCh38
rs938697037 Chr.2: 9630027-9630027 on Build GRCh38
rs904356102 Chr.2: 9629905-9629905 on Build GRCh38
rs369055092 Chr.2: 9630144-9630144 on Build GRCh38
rs904367772 Chr.2: 9630071-9630071 on Build GRCh38
rs372545955 Chr.2: 9630182-9630182 on Build GRCh38
rs535335896 Chr.2: 9630041-9630041 on Build GRCh38
rs1042907982 Chr.2: 9629902-9629902 on Build GRCh38
rs757395602 Chr.2: 9630246-9630246 on Build GRCh38
rs748667930 Chr.2: 9630110-9630110 on Build GRCh38
rs758380692 Chr.2: 9630362-9630362 on Build GRCh38
rs542147048 Chr.2: 9629952-9629952 on Build GRCh38
rs1027235223 Chr.2: 9630061-9630062 on Build GRCh38
rs761396781 Chr.2: 9630169-9630169 on Build GRCh38
rs973559819 Chr.2: 9629978-9629978 on Build GRCh38
rs570403921 Chr.2: 9629965-9629965 on Build GRCh38
rs368512939 Chr.2: 9630137-9630137 on Build GRCh38
rs945393867 Chr.2: 9629951-9629951 on Build GRCh38
rs760149820 Chr.2: 9630328-9630328 on Build GRCh38
rs1043744634 Chr.2: 9629952-9629952 on Build GRCh38
rs781552947 Chr.2: 9630113-9630113 on Build GRCh38
rs1009212767 Chr.2: 9630094-9630094 on Build GRCh38
rs993160517 Chr.2: 9630049-9630049 on Build GRCh38
rs1021090213 Chr.2: 9630097-9630098 on Build GRCh38

View Full Product Details