Product Details

Assay Reference Genome
Location

Chr.1:17000029 on build GRCh38
Cytoband
1p36.13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23400
Gene Symbol
ATP13A2
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Location
Chr.1:16985958-17011972 on build GRCh38
Assay Gene Location
Overlaps Intron 11 - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ATP13A2 NM_001141973.2 NP_001135445.1
NM_001141974.2 NP_001135446.1
NM_022089.3 NP_071372.1
XM_005245810.1 XP_005245867.1
XM_005245811.1 XP_005245868.1
XM_005245812.1 XP_005245869.1
XM_005245815.1 XP_005245872.1
XM_006710512.1 XP_006710575.1
XM_006710513.1 XP_006710576.1
XM_011541128.1 XP_011539430.1
XM_011541129.1 XP_011539431.1
XM_017000844.1 XP_016856333.1
XM_017000845.1 XP_016856334.1
XM_017000846.1 XP_016856335.1
XM_017000847.1 XP_016856336.1
XM_017000848.1 XP_016856337.1
XM_017000849.1 XP_016856338.1
XM_017000850.1 XP_016856339.1
AK075310.1 BAC11539.1
AK290210.1
AL354615.1 CAB89728.1
AY461712.1 AAR23423.1
AY987009.1 AAX88804.1
BC030267.1 AAH30267.1
BC034575.1 AAH34575.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv508947 Chr1:16909817 - 17011115 on Build GRCh38 Insertion MFAP2 ATP13A2 CROCC
nsv954854 Chr1:16961106 - 17013905 on Build GRCh38 Deletion MFAP2 ATP13A2 CROCC
nsv428421 Chr1:16437838 - 17157486 on Build GRCh38 Gain+Loss PADI2 SPATA21 RNU1-1 RNU1-4 MIR3675 RNU1-3 LOC102723383 CROCC MST1P2 MST1L SDHB LOC105376796 LOC105376805 FAM231B NECAP2 FAM231A ESPNP RNU1-2 LOC100132147 MFAP2 CROCCP3 CROCCP2 NBPF1 ATP13A2
dgv108n100 Chr1:16850538 - 17010370 on Build GRCh38 Gain LOC105376805 MIR3675 RNU1-2 MFAP2 ATP13A2 CROCC
dgv92n100 Chr1:16684943 - 17003642 on Build GRCh38 Gain MST1L LOC105376805 ESPNP RNU1-4 MIR3675 RNU1-2 MFAP2 ATP13A2 LOC102723383 CROCC
nsv834280 Chr1:16888218 - 17123006 on Build GRCh38 Loss PADI2 SDHB LOC105376805 RNU1-2 MFAP2 ATP13A2 CROCC
nsv834269 Chr1:16855283 - 17038531 on Build GRCh38 Loss SDHB LOC105376805 MIR3675 RNU1-2 MFAP2 ATP13A2 CROCC

View Full Product Details