Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77210234-77210773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3217985 c.506G>A p.R169Q Chr.9: 77210626-77210626 on Build GRCh38
3217984 c.506G>A p.R169Q Chr.9: 77210626-77210626 on Build GRCh38
7287278 c.496-2A>C p.? Chr.9: 77210614-77210614 on Build GRCh38
6903139 c.507G>A p.R169R Chr.9: 77210627-77210627 on Build GRCh38
6903141 c.507G>A p.R169R Chr.9: 77210627-77210627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781084069 Chr.9: 77210726-77210726 on Build GRCh38
rs879199752 Chr.9: 77210695-77210695 on Build GRCh38
rs972967614 Chr.9: 77210332-77210332 on Build GRCh38
rs1000705098 Chr.9: 77210295-77210295 on Build GRCh38
rs1042276761 Chr.9: 77210283-77210283 on Build GRCh38
rs766587340 Chr.9: 77210639-77210639 on Build GRCh38
rs753300394 Chr.9: 77210679-77210679 on Build GRCh38
rs752813760 Chr.9: 77210568-77210568 on Build GRCh38
rs141109675 Chr.9: 77210636-77210636 on Build GRCh38
rs117509387 Chr.9: 77210352-77210352 on Build GRCh38
rs139380985 Chr.9: 77210625-77210625 on Build GRCh38
rs909788090 Chr.9: 77210421-77210421 on Build GRCh38
rs928077925 Chr.9: 77210346-77210346 on Build GRCh38
rs544104609 Chr.9: 77210608-77210608 on Build GRCh38
rs188628135 Chr.9: 77210310-77210310 on Build GRCh38
rs377104687 Chr.9: 77210589-77210589 on Build GRCh38
rs919217389 Chr.9: 77210386-77210386 on Build GRCh38
rs768517006 Chr.9: 77210694-77210694 on Build GRCh38
rs774869789 Chr.9: 77210651-77210651 on Build GRCh38
rs535082078 Chr.9: 77210401-77210401 on Build GRCh38
rs369548882 Chr.9: 77210727-77210727 on Build GRCh38
rs756330178 Chr.9: 77210569-77210569 on Build GRCh38
rs961331026 Chr.9: 77210320-77210320 on Build GRCh38
rs546023010 Chr.9: 77210716-77210716 on Build GRCh38
rs1008208191 Chr.9: 77210308-77210308 on Build GRCh38
rs769393043 Chr.9: 77210626-77210626 on Build GRCh38
rs780209183 Chr.9: 77210539-77210539 on Build GRCh38
rs779057916 Chr.9: 77210704-77210704 on Build GRCh38
rs768562676 Chr.9: 77210613-77210613 on Build GRCh38
rs942533402 Chr.9: 77210426-77210426 on Build GRCh38
rs371449823 Chr.9: 77210717-77210717 on Build GRCh38
rs760037642 Chr.9: 77210654-77210654 on Build GRCh38
rs10869911 Chr.9: 77210368-77210368 on Build GRCh38
rs142041147 Chr.9: 77210554-77210554 on Build GRCh38
rs1027682562 Chr.9: 77210735-77210735 on Build GRCh38
rs952979414 Chr.9: 77210741-77210741 on Build GRCh38
rs1019591815 Chr.9: 77210314-77210314 on Build GRCh38
rs772689261 Chr.9: 77210635-77210635 on Build GRCh38
rs953032049 Chr.9: 77210450-77210450 on Build GRCh38
rs972083958 Chr.9: 77210653-77210653 on Build GRCh38
rs370541609 Chr.9: 77210607-77210607 on Build GRCh38
rs960861350 Chr.9: 77210355-77210355 on Build GRCh38
rs1039533963 Chr.9: 77210455-77210455 on Build GRCh38
rs768135367 Chr.9: 77210666-77210666 on Build GRCh38
rs1027198425 Chr.9: 77210445-77210445 on Build GRCh38
rs889866532 Chr.9: 77210304-77210304 on Build GRCh38
rs1028862705 Chr.9: 77210303-77210303 on Build GRCh38
rs903690683 Chr.9: 77210285-77210285 on Build GRCh38
rs757482674 Chr.9: 77210709-77210709 on Build GRCh38
rs900934485 Chr.9: 77210474-77210474 on Build GRCh38
rs764115216 Chr.9: 77210599-77210602 on Build GRCh38
rs146334644 Chr.9: 77210696-77210696 on Build GRCh38
rs756128729 Chr.9: 77210687-77210687 on Build GRCh38
rs1048824899 Chr.9: 77210421-77210422 on Build GRCh38
rs367807671 Chr.9: 77210693-77210693 on Build GRCh38
rs991883080 Chr.9: 77210556-77210556 on Build GRCh38
rs764260920 Chr.9: 77210689-77210689 on Build GRCh38
rs756970615 Chr.9: 77210715-77210715 on Build GRCh38
rs945292311 Chr.9: 77210272-77210272 on Build GRCh38
rs549934747 Chr.9: 77210530-77210530 on Build GRCh38
rs751582738 Chr.9: 77210709-77210710 on Build GRCh38
rs997899564 Chr.9: 77210542-77210542 on Build GRCh38

View Full Product Details