Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 22056387-22056893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
UniGene
Hs.712707
Gene Chromosome Location
Chr.6: 21666444-22194400 on Build GRCh38
Transcripts
NR_015410.1

SNP Detail

SNP ID Chromosome Location
rs914409408 Chr.6: 22056626-22056626 on Build GRCh38
rs543073579 Chr.6: 22056592-22056592 on Build GRCh38
rs766303274 Chr.6: 22056593-22056593 on Build GRCh38
rs532905380 Chr.6: 22056675-22056675 on Build GRCh38
rs1031751705 Chr.6: 22056491-22056491 on Build GRCh38
rs1008152921 Chr.6: 22056742-22056742 on Build GRCh38
rs896053750 Chr.6: 22056433-22056433 on Build GRCh38
rs1038513968 Chr.6: 22056711-22056711 on Build GRCh38
rs373187363 Chr.6: 22056506-22056506 on Build GRCh38
rs986788172 Chr.6: 22056563-22056563 on Build GRCh38
rs149073114 Chr.6: 22056619-22056619 on Build GRCh38
rs369931606 Chr.6: 22056702-22056702 on Build GRCh38
rs189359572 Chr.6: 22056500-22056500 on Build GRCh38
rs530250464 Chr.6: 22056710-22056710 on Build GRCh38
rs566454042 Chr.6: 22056743-22056743 on Build GRCh38
rs955969146 Chr.6: 22056617-22056617 on Build GRCh38
rs898655862 Chr.6: 22056718-22056718 on Build GRCh38
rs764064589 Chr.6: 22056723-22056723 on Build GRCh38
rs569480982 Chr.6: 22056798-22056798 on Build GRCh38
rs929485145 Chr.6: 22056419-22056419 on Build GRCh38
rs373938791 Chr.6: 22056633-22056633 on Build GRCh38
rs550684813 Chr.6: 22056721-22056721 on Build GRCh38
rs766332812 Chr.6: 22056591-22056591 on Build GRCh38
rs969912797 Chr.6: 22056815-22056815 on Build GRCh38
rs537646015 Chr.6: 22056455-22056455 on Build GRCh38
rs948516097 Chr.6: 22056454-22056454 on Build GRCh38
rs1047443935 Chr.6: 22056679-22056679 on Build GRCh38
rs1053640750 Chr.6: 22056753-22056753 on Build GRCh38
rs751597273 Chr.6: 22056747-22056747 on Build GRCh38
rs2294214 Chr.6: 22056694-22056694 on Build GRCh38
rs1028861057 Chr.6: 22056518-22056518 on Build GRCh38
rs996949275 Chr.6: 22056516-22056516 on Build GRCh38
rs898537721 Chr.6: 22056513-22056513 on Build GRCh38
rs955399529 Chr.6: 22056523-22056523 on Build GRCh38
rs369186301 Chr.6: 22056515-22056515 on Build GRCh38
rs1004356911 Chr.6: 22056502-22056502 on Build GRCh38
rs551800989 Chr.6: 22056835-22056835 on Build GRCh38
rs540643056 Chr.6: 22056647-22056647 on Build GRCh38
rs980365102 Chr.6: 22056846-22056846 on Build GRCh38
rs904612216 Chr.6: 22056463-22056463 on Build GRCh38
rs938379736 Chr.6: 22056480-22056480 on Build GRCh38
rs575635252 Chr.6: 22056512-22056512 on Build GRCh38
rs1056726985 Chr.6: 22056489-22056489 on Build GRCh38
rs979865629 Chr.6: 22056658-22056658 on Build GRCh38
rs891233803 Chr.6: 22056738-22056738 on Build GRCh38
rs904558963 Chr.6: 22056731-22056731 on Build GRCh38
rs2294215 Chr.6: 22056453-22056453 on Build GRCh38
rs906873482 Chr.6: 22056476-22056476 on Build GRCh38
rs568197438 Chr.6: 22056634-22056634 on Build GRCh38
rs963075145 Chr.6: 22056627-22056627 on Build GRCh38
rs954604063 Chr.6: 22056575-22056575 on Build GRCh38
rs36015120 Chr.6: 22056413-22056414 on Build GRCh38
rs760410620 Chr.6: 22056469-22056469 on Build GRCh38
rs527269611 Chr.6: 22056765-22056765 on Build GRCh38
rs997462896 Chr.6: 22056720-22056720 on Build GRCh38
rs565570719 Chr.6: 22056668-22056668 on Build GRCh38
rs990030903 Chr.6: 22056781-22056781 on Build GRCh38
rs1054244977 Chr.6: 22056413-22056413 on Build GRCh38
rs1057413789 Chr.6: 22056427-22056427 on Build GRCh38

View Full Product Details