Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 22135901-22136410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC15
Entrez Gene ID
401237
Gene Name
cancer susceptibility candidate 15 (non-protein coding)
Gene Aliases
LINC00340
UniGene
Hs.712707
Gene Chromosome Location
Chr.6: 21666444-22194400 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

Gene Symbol
NBAT1
Entrez Gene ID
729177
Gene Name
neuroblastoma associated transcript 1
Gene Aliases
CASC14, NBAT-1
UniGene
Hs.730673
Gene Chromosome Location
Chr.6: 22134602-22147193 on Build GRCh38
Transcripts
NR_015410.1 NR_034143.1

SNP Detail

SNP ID Chromosome Location
rs927195860 Chr.6: 22136186-22136186 on Build GRCh38
rs1054103851 Chr.6: 22136213-22136213 on Build GRCh38
rs1005448282 Chr.6: 22136293-22136293 on Build GRCh38
rs892776106 Chr.6: 22136228-22136228 on Build GRCh38
rs775163866 Chr.6: 22136338-22136338 on Build GRCh38
rs906611193 Chr.6: 22136236-22136236 on Build GRCh38
rs1029833996 Chr.6: 22136087-22136087 on Build GRCh38
rs1047084681 Chr.6: 22136261-22136261 on Build GRCh38
rs1025474426 Chr.6: 22135952-22135952 on Build GRCh38
rs4712656 Chr.6: 22136033-22136033 on Build GRCh38
rs888350020 Chr.6: 22136262-22136262 on Build GRCh38
rs538356667 Chr.6: 22136163-22136163 on Build GRCh38
rs9466269 Chr.6: 22136180-22136180 on Build GRCh38
rs566566401 Chr.6: 22136248-22136248 on Build GRCh38
rs920020260 Chr.6: 22136150-22136150 on Build GRCh38
rs6915322 Chr.6: 22135931-22135931 on Build GRCh38
rs916732746 Chr.6: 22136181-22136181 on Build GRCh38
rs184847896 Chr.6: 22136337-22136337 on Build GRCh38
rs148309004 Chr.6: 22135947-22135947 on Build GRCh38
rs891340280 Chr.6: 22136274-22136274 on Build GRCh38
rs1046439904 Chr.6: 22136223-22136223 on Build GRCh38
rs1018525944 Chr.6: 22136035-22136035 on Build GRCh38
rs1018950995 Chr.6: 22136294-22136294 on Build GRCh38
rs551223297 Chr.6: 22136102-22136102 on Build GRCh38
rs967032160 Chr.6: 22136121-22136121 on Build GRCh38
rs1029411495 Chr.6: 22136387-22136387 on Build GRCh38
rs552307078 Chr.6: 22136331-22136331 on Build GRCh38
rs768434386 Chr.6: 22136314-22136314 on Build GRCh38
rs892857926 Chr.6: 22135946-22135946 on Build GRCh38
rs976505440 Chr.6: 22136075-22136075 on Build GRCh38
rs937013855 Chr.6: 22136193-22136193 on Build GRCh38
rs548837763 Chr.6: 22136187-22136187 on Build GRCh38
rs1045625273 Chr.6: 22136184-22136184 on Build GRCh38
rs6915330 Chr.6: 22135943-22135943 on Build GRCh38
rs924182985 Chr.6: 22136160-22136160 on Build GRCh38
rs747421706 Chr.6: 22136117-22136117 on Build GRCh38
rs1005332329 Chr.6: 22136015-22136015 on Build GRCh38
rs998429361 Chr.6: 22136382-22136382 on Build GRCh38
rs888402270 Chr.6: 22135970-22135970 on Build GRCh38
rs955510404 Chr.6: 22135987-22135987 on Build GRCh38
rs187478474 Chr.6: 22135930-22135930 on Build GRCh38
rs192440368 Chr.6: 22136314-22136314 on Build GRCh38
rs74453603 Chr.6: 22136134-22136134 on Build GRCh38
rs532412802 Chr.6: 22136044-22136044 on Build GRCh38
rs1041478989 Chr.6: 22136273-22136273 on Build GRCh38

View Full Product Details