Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144511678-144511825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982726 c.3497G>T p.G1166V Chr.8: 144511686-144511686 on Build GRCh38
3834300 c.3447G>T p.L1149L Chr.8: 144511736-144511736 on Build GRCh38
6958348 c.3474G>T p.R1158S Chr.8: 144511709-144511709 on Build GRCh38
6951617 c.3424G>A p.D1142N Chr.8: 144511759-144511759 on Build GRCh38
5019569 c.3465C>T p.F1155F Chr.8: 144511718-144511718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752301125 Chr.8: 144511790-144511790 on Build GRCh38
rs202134651 Chr.8: 144511753-144511753 on Build GRCh38
rs587778647 Chr.8: 144511762-144511762 on Build GRCh38
rs529113837 Chr.8: 144511754-144511754 on Build GRCh38
rs950715422 Chr.8: 144511773-144511773 on Build GRCh38
rs539751764 Chr.8: 144511802-144511802 on Build GRCh38
rs780676589 Chr.8: 144511734-144511734 on Build GRCh38
rs780986647 Chr.8: 144511724-144511726 on Build GRCh38
rs778513153 Chr.8: 144511784-144511784 on Build GRCh38
rs769772821 Chr.8: 144511803-144511803 on Build GRCh38
rs34914400 Chr.8: 144511703-144511703 on Build GRCh38
rs747815189 Chr.8: 144511698-144511698 on Build GRCh38
rs747758347 Chr.8: 144511747-144511747 on Build GRCh38
rs562024579 Chr.8: 144511751-144511751 on Build GRCh38
rs757502009 Chr.8: 144511720-144511720 on Build GRCh38
rs35346077 Chr.8: 144511740-144511740 on Build GRCh38
rs371037028 Chr.8: 144511781-144511781 on Build GRCh38
rs758089563 Chr.8: 144511791-144511791 on Build GRCh38
rs916484589 Chr.8: 144511702-144511702 on Build GRCh38
rs767822615 Chr.8: 144511764-144511764 on Build GRCh38
rs763080171 Chr.8: 144511715-144511715 on Build GRCh38
rs773088003 Chr.8: 144511699-144511699 on Build GRCh38
rs774386060 Chr.8: 144511805-144511805 on Build GRCh38
rs748732922 Chr.8: 144511738-144511738 on Build GRCh38
rs376874393 Chr.8: 144511718-144511718 on Build GRCh38
rs370611564 Chr.8: 144511760-144511760 on Build GRCh38
rs777542762 Chr.8: 144511793-144511793 on Build GRCh38
rs61755066 Chr.8: 144511748-144511748 on Build GRCh38
rs61755067 Chr.8: 144511759-144511759 on Build GRCh38
rs775431574 Chr.8: 144511710-144511710 on Build GRCh38
rs770697720 Chr.8: 144511799-144511799 on Build GRCh38
rs564143545 Chr.8: 144511726-144511726 on Build GRCh38
rs746892308 Chr.8: 144511752-144511752 on Build GRCh38
rs756549605 Chr.8: 144511727-144511727 on Build GRCh38
rs375432640 Chr.8: 144511765-144511765 on Build GRCh38
rs767821367 Chr.8: 144511722-144511722 on Build GRCh38
rs761157177 Chr.8: 144511766-144511766 on Build GRCh38
rs759478327 Chr.8: 144511755-144511755 on Build GRCh38
rs766385605 Chr.8: 144511707-144511707 on Build GRCh38
rs370677156 Chr.8: 144511800-144511800 on Build GRCh38
rs1043628799 Chr.8: 144511708-144511708 on Build GRCh38
rs746656592 Chr.8: 144511798-144511798 on Build GRCh38
rs754479731 Chr.8: 144511739-144511739 on Build GRCh38
rs754328779 Chr.8: 144511772-144511772 on Build GRCh38
rs373652049 Chr.8: 144511716-144511716 on Build GRCh38

View Full Product Details