Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144511402-144511575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6929290 c.3619G>A p.A1207T Chr.8: 144511439-144511439 on Build GRCh38
6764693 c.3600G>A p.T1200T Chr.8: 144511458-144511458 on Build GRCh38
5438769 c.3551G>C p.W1184S Chr.8: 144511507-144511507 on Build GRCh38
4490272 c.3610C>T p.L1204L Chr.8: 144511448-144511448 on Build GRCh38
6764694 c.3623G>A p.R1208H Chr.8: 144511435-144511435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587778648 Chr.8: 144511537-144511537 on Build GRCh38
rs567937389 Chr.8: 144511491-144511491 on Build GRCh38
rs774756135 Chr.8: 144511504-144511504 on Build GRCh38
rs781313959 Chr.8: 144511456-144511456 on Build GRCh38
rs757292188 Chr.8: 144511533-144511533 on Build GRCh38
rs778583430 Chr.8: 144511441-144511441 on Build GRCh38
rs370069034 Chr.8: 144511516-144511516 on Build GRCh38
rs751477241 Chr.8: 144511532-144511532 on Build GRCh38
rs35225938 Chr.8: 144511458-144511458 on Build GRCh38
rs758188100 Chr.8: 144511529-144511529 on Build GRCh38
rs754019797 Chr.8: 144511539-144511539 on Build GRCh38
rs386833854 Chr.8: 144511458-144511459 on Build GRCh38
rs41555416 Chr.8: 144511436-144511436 on Build GRCh38
rs546643758 Chr.8: 144511468-144511468 on Build GRCh38
rs776146178 Chr.8: 144511526-144511526 on Build GRCh38
rs772747279 Chr.8: 144511476-144511476 on Build GRCh38
rs955049317 Chr.8: 144511444-144511444 on Build GRCh38
rs939186872 Chr.8: 144511522-144511522 on Build GRCh38
rs376981214 Chr.8: 144511514-144511514 on Build GRCh38
rs563816306 Chr.8: 144511435-144511435 on Build GRCh38
rs752487649 Chr.8: 144511489-144511489 on Build GRCh38
rs879192998 Chr.8: 144511535-144511535 on Build GRCh38
rs1005731353 Chr.8: 144511548-144511548 on Build GRCh38
rs779070043 Chr.8: 144511543-144511543 on Build GRCh38
rs745722387 Chr.8: 144511501-144511501 on Build GRCh38
rs752292609 Chr.8: 144511421-144511421 on Build GRCh38
rs762230834 Chr.8: 144511428-144511428 on Build GRCh38
rs56341125 Chr.8: 144511434-144511434 on Build GRCh38
rs201809411 Chr.8: 144511478-144511478 on Build GRCh38
rs139228543 Chr.8: 144511527-144511527 on Build GRCh38
rs764685694 Chr.8: 144511425-144511425 on Build GRCh38
rs774611413 Chr.8: 144511470-144511470 on Build GRCh38
rs767956471 Chr.8: 144511473-144511473 on Build GRCh38
rs772265082 Chr.8: 144511549-144511549 on Build GRCh38
rs765036646 Chr.8: 144511488-144511488 on Build GRCh38
rs371461701 Chr.8: 144511472-144511472 on Build GRCh38
rs748529710 Chr.8: 144511546-144511546 on Build GRCh38
rs768782202 Chr.8: 144511497-144511497 on Build GRCh38
rs1038354357 Chr.8: 144511550-144511550 on Build GRCh38
rs770138542 Chr.8: 144511459-144511459 on Build GRCh38
rs780285435 Chr.8: 144511496-144511496 on Build GRCh38
rs557256260 Chr.8: 144511513-144511513 on Build GRCh38
rs753427189 Chr.8: 144511480-144511480 on Build GRCh38
rs530184653 Chr.8: 144511426-144511426 on Build GRCh38
rs55805048 Chr.8: 144511422-144511422 on Build GRCh38
rs765804620 Chr.8: 144511517-144511517 on Build GRCh38
rs367898813 Chr.8: 144511534-144511534 on Build GRCh38
rs758966528 Chr.8: 144511461-144511484 on Build GRCh38
rs745930478 Chr.8: 144511493-144511493 on Build GRCh38
rs146398243 Chr.8: 144511464-144511464 on Build GRCh38
rs763479657 Chr.8: 144511423-144511423 on Build GRCh38
rs201384843 Chr.8: 144511449-144511449 on Build GRCh38
rs754822264 Chr.8: 144511440-144511440 on Build GRCh38
rs763965257 Chr.8: 144511530-144511530 on Build GRCh38
rs748047415 Chr.8: 144511445-144511445 on Build GRCh38
rs199885535 Chr.8: 144511485-144511485 on Build GRCh38
rs535259922 Chr.8: 144511495-144511495 on Build GRCh38
rs775914502 Chr.8: 144511461-144511461 on Build GRCh38
rs370738615 Chr.8: 144511551-144511551 on Build GRCh38

View Full Product Details