Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51047089-51047248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
7212309 c.188delC p.T63fs*31 Chr.18: 51047234-51047234 on Build GRCh38
189728 c.97G>T p.E33* Chr.18: 51047143-51047143 on Build GRCh38
30786 c.74G>A p.C25Y Chr.18: 51047120-51047120 on Build GRCh38
6929188 c.91_102>TT p.E31fs*11 Chr.18: 51047137-51047148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019245952 Chr.18: 51047160-51047160 on Build GRCh38
rs758408642 Chr.18: 51047161-51047161 on Build GRCh38
rs786202127 Chr.18: 51047150-51047150 on Build GRCh38
rs876660712 Chr.18: 51047199-51047199 on Build GRCh38
rs786203560 Chr.18: 51047199-51047200 on Build GRCh38
rs786204166 Chr.18: 51047218-51047218 on Build GRCh38
rs939612648 Chr.18: 51047165-51047165 on Build GRCh38
rs749503989 Chr.18: 51047196-51047196 on Build GRCh38
rs146104321 Chr.18: 51047148-51047148 on Build GRCh38
rs878854764 Chr.18: 51047164-51047164 on Build GRCh38
rs965942065 Chr.18: 51047178-51047178 on Build GRCh38
rs863224731 Chr.18: 51047173-51047173 on Build GRCh38
rs778465458 Chr.18: 51047130-51047130 on Build GRCh38
rs876660353 Chr.18: 51047136-51047136 on Build GRCh38
rs886053893 Chr.18: 51047133-51047133 on Build GRCh38
rs780090544 Chr.18: 51047192-51047192 on Build GRCh38
rs746732669 Chr.18: 51047176-51047176 on Build GRCh38
rs770789755 Chr.18: 51047185-51047185 on Build GRCh38
rs777917225 Chr.18: 51047163-51047163 on Build GRCh38
rs587781977 Chr.18: 51047221-51047221 on Build GRCh38
rs768796731 Chr.18: 51047205-51047205 on Build GRCh38
rs1053158497 Chr.18: 51047206-51047206 on Build GRCh38
rs876659391 Chr.18: 51047116-51047116 on Build GRCh38

View Full Product Details