Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51048708-51048871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6957607 c.308A>C p.D103A Chr.18: 51048744-51048744 on Build GRCh38
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
4382624 c.290G>A p.R97H Chr.18: 51048726-51048726 on Build GRCh38
7430437 c.402A>T p.E134D Chr.18: 51048838-51048838 on Build GRCh38
5521010 c.275A>T p.H92L Chr.18: 51048711-51048711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202126703 Chr.18: 51048730-51048730 on Build GRCh38
rs779069779 Chr.18: 51048835-51048835 on Build GRCh38
rs140926102 Chr.18: 51048805-51048805 on Build GRCh38
rs755862230 Chr.18: 51048826-51048826 on Build GRCh38
rs748395067 Chr.18: 51048836-51048836 on Build GRCh38
rs876659844 Chr.18: 51048780-51048780 on Build GRCh38
rs727504151 Chr.18: 51048762-51048762 on Build GRCh38
rs377767325 Chr.18: 51048817-51048818 on Build GRCh38
rs863224401 Chr.18: 51048811-51048811 on Build GRCh38
rs771808272 Chr.18: 51048810-51048810 on Build GRCh38
rs751154230 Chr.18: 51048734-51048734 on Build GRCh38
rs377767324 Chr.18: 51048809-51048810 on Build GRCh38
rs750172880 Chr.18: 51048807-51048807 on Build GRCh38
rs754123998 Chr.18: 51048777-51048778 on Build GRCh38
rs377767323 Chr.18: 51048738-51048738 on Build GRCh38
rs145988618 Chr.18: 51048790-51048790 on Build GRCh38
rs150229208 Chr.18: 51048823-51048823 on Build GRCh38
rs876660079 Chr.18: 51048733-51048733 on Build GRCh38
rs757211048 Chr.18: 51048778-51048778 on Build GRCh38
rs377767326 Chr.18: 51048839-51048839 on Build GRCh38

View Full Product Details