Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51054795-51054956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
6965857 c.542C>T p.T181I Chr.18: 51054868-51054868 on Build GRCh38
5030960 c.527G>A p.G176E Chr.18: 51054853-51054853 on Build GRCh38
6438227 c.623C>T p.A208V Chr.18: 51054949-51054949 on Build GRCh38
14157 c.607_608delCC p.P203fs*2 Chr.18: 51054933-51054934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946539148 Chr.18: 51054912-51054912 on Build GRCh38
rs377767330 Chr.18: 51054842-51054853 on Build GRCh38
rs778576111 Chr.18: 51054845-51054845 on Build GRCh38
rs61751988 Chr.18: 51054894-51054894 on Build GRCh38
rs368528856 Chr.18: 51054851-51054851 on Build GRCh38
rs760236686 Chr.18: 51054877-51054877 on Build GRCh38
rs587780792 Chr.18: 51054901-51054901 on Build GRCh38
rs752575871 Chr.18: 51054898-51054898 on Build GRCh38
rs199809905 Chr.18: 51054933-51054933 on Build GRCh38
rs876659426 Chr.18: 51054833-51054833 on Build GRCh38
rs140743238 Chr.18: 51054891-51054891 on Build GRCh38
rs145805120 Chr.18: 51054908-51054908 on Build GRCh38
rs759288477 Chr.18: 51054892-51054892 on Build GRCh38
rs927620013 Chr.18: 51054886-51054886 on Build GRCh38
rs542392980 Chr.18: 51054861-51054861 on Build GRCh38
rs786203737 Chr.18: 51054925-51054925 on Build GRCh38
rs200717327 Chr.18: 51054896-51054896 on Build GRCh38
rs778555013 Chr.18: 51054827-51054828 on Build GRCh38
rs138800446 Chr.18: 51054847-51054847 on Build GRCh38
rs377767331 Chr.18: 51054859-51054859 on Build GRCh38
rs761936246 Chr.18: 51054899-51054899 on Build GRCh38
rs547278031 Chr.18: 51054920-51054920 on Build GRCh38
rs144226135 Chr.18: 51054836-51054836 on Build GRCh38
rs748018891 Chr.18: 51054849-51054849 on Build GRCh38
rs770798845 Chr.18: 51054880-51054880 on Build GRCh38
rs377767332 Chr.18: 51054864-51054864 on Build GRCh38
rs780665234 Chr.18: 51054932-51054932 on Build GRCh38

View Full Product Details