Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8orf33
Entrez Gene ID
65265
Gene Name
chromosome 8 open reading frame 33
Gene Aliases
-
Chromosome Location
Chr.8: 145052438 - 145056030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023080.2 NP_075568.1 3 - 4 457 73
GenBank mRNA AK024642.1 - 3 - 4 455 73
BC010001.2 - 3 - 4 417 73
BC015181.1 - 2 - 3 218 73
BX648996.1 - 2 - 3 517 73
CR457330.1 - 2 - 3 403 73
DA467518.1 - 3 - 4 457 73

View Full Product Details