Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
Chromosome Location
Chr.5: 181039875 - 181061525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK057097.1 - 6 - 7 1866 100

View Full Product Details