Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C9orf66
Entrez Gene ID
157983
Gene Name
chromosome 9 open reading frame 66
Gene Aliases
-
Chromosome Location
Chr.9: 213108 - 215893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152569.2 NP_689782.2 1 - 1 701 120
GenBank mRNA AK055720.1 - 1 - 1 549 120

View Full Product Details