Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC57
- Entrez Gene ID
-
284001
- Gene Name
- coiled-coil domain containing 57
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 82101458 - 82212892 on Build GRCh38
View Full Product Details