The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016955.3 | NP_058651.3 | 1 - 2 | 332 | 63 |
XM_017008278.1 | XP_016863767.1 | 1 - 2 | 283 | 63 | |
GenBank mRNA | AJ238617.1 | - | 1 - 2 | 113 | 63 |
AK292476.1 | - | 1 - 2 | 332 | 63 | |
BC126213.1 | - | 1 - 2 | 39 | 63 | |
BX648976.1 | - | 1 - 2 | 193 | 63 |