The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172000.3 | NP_741997.3 | 1 - 1 | 2264 | 83 |
GenBank mRNA | AJ515384.2 | - | 1 - 1 | 2257 | 83 |
GQ472227.1 | - | 1 - 1 | 2257 | 83 |