Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC30
Entrez Gene ID
84277
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
Gene Aliases
WBSCR18
Chromosome Location
Chr.7: 73680918 - 73683451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032317.2 NP_115693.2 1 - 1 352 91
GenBank mRNA AF412025.1 - 1 - 1 343 91
AK058113.1 - 1 - 1 332 91
AK094305.1 - 1 - 1 352 91
BC005056.1 - 1 - 1 343 91

View Full Product Details