The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201484.1 | NP_001188413.1 | 10 - 11 | 2101 | 88 |
NM_032634.3 | NP_116023.2 | 8 - 9 | 3254 | 88 | |
NM_152850.3 | NP_690577.2 | 9 - 10 | 1649 | 88 | |
XM_005251619.3 | XP_005251676.1 | 8 - 9 | 2879 | 88 | |
XM_017015222.1 | XP_016870711.1 | 9 - 10 | 3371 | 88 | |
XM_017015223.1 | XP_016870712.1 | 9 - 10 | 2030 | 88 | |
XM_017015224.1 | XP_016870713.1 | 9 - 10 | 2294 | 88 | |
GenBank mRNA | AL833956.1 | - | 8 - 9 | 3287 | 88 |
AY358472.1 | - | 7 - 8 | 2860 | 88 | |
BC001030.2 | - | 6 - 7 | 979 | 88 | |
BC013987.2 | - | 7 - 8 | 1065 | 88 | |
BC029271.1 | - | 7 - 8 | 2525 | 88 | |
BC036916.1 | - | 8 - 9 | 1614 | 88 | |
BC065282.1 | - | 10 - 11 | 1772 | 88 |