The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172238.3 | NP_758438.2 | 3 - 4 | 1111 | 78 |
GenBank mRNA | AY028376.1 | - | 3 - 4 | 1111 | 78 |