Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHZ2
Entrez Gene ID
128553
Gene Name
teashirt zinc finger homeobox 2
Gene Aliases
C20orf17, OVC10-2, TSH2, ZABC2, ZNF218
Chromosome Location
Chr.20: 52972309 - 53495330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193421.1 NP_001180350.1 2 - 3 3466 71
NM_173485.5 NP_775756.3 2 - 3 3999 71
GenBank mRNA AK091206.1 - 2 - 3 3192 71
AK302570.1 - 2 - 3 3466 71
BX640770.1 - 2 - 3 3999 71
BX648148.1 - 1 - 2 1434 71

View Full Product Details