The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164673.1 | NP_001158145.1 | 6 - 7 | 830 | 107 |
NM_001256896.1 | NP_001243825.1 | 3 - 4 | 539 | 107 | |
NM_001301071.1 | NP_001288000.1 | 6 - 7 | 841 | 107 | |
NM_173660.4 | NP_775931.3 | 6 - 7 | 841 | 107 | |
XM_011513435.2 | XP_011511737.1 | 6 - 7 | 841 | 107 | |
XM_011513437.2 | XP_011511739.1 | 6 - 7 | 830 | 107 | |
XM_017008038.1 | XP_016863527.1 | 4 - 5 | 404 | 107 | |
GenBank mRNA | AB220918.1 | - | 6 - 7 | 771 | 107 |
AK091037.1 | - | 6 - 7 | 825 | 107 | |
AK096456.1 | - | 3 - 4 | 539 | 107 | |
BC043568.1 | - | 2 - 3 | 178 | 107 | |
BC062369.1 | - | 2 - 3 | 233 | 107 | |
BC131544.1 | - | 6 - 7 | 778 | 107 | |
BC141852.1 | - | 6 - 7 | 812 | 107 |