Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
Chromosome Location
Chr.22: 26443613 - 26483863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022081.5 NP_071364.4 4 - 5 941 109
NM_152841.2 NP_690054.1 2 - 3 558 109
NR_073135.1 4 - 5 850 109
NR_073136.1 2 - 3 558 109
XM_006724353.3 XP_006724416.1 4 - 5 932 109
XM_006724354.3 XP_006724417.1 4 - 5 837 109
XM_011530485.1 XP_011528787.1 4 - 5 932 109
XM_011530486.2 XP_011528788.1 4 - 5 837 109
XM_011530487.2 XP_011528789.1 5 - 6 1042 109
XM_011530488.2 XP_011528790.1 4 - 5 840 109
XM_011530489.2 XP_011528791.1 5 - 6 943 109
XM_011530490.2 XP_011528792.1 4 - 5 932 109
XM_011530491.2 XP_011528793.1 4 - 5 932 109
XM_011530492.1 XP_011528794.1 4 - 5 932 109
XM_011530493.2 XP_011528795.1 4 - 5 932 109
XM_011530494.2 XP_011528796.1 2 - 3 228 109
XM_017029042.1 XP_016884531.1 4 - 5 837 109
XM_017029043.1 XP_016884532.1 5 - 6 1042 109
XM_017029044.1 XP_016884533.1 4 - 5 840 109
XM_017029045.1 XP_016884534.1 4 - 5 932 109
XM_017029046.1 XP_016884535.1 4 - 5 932 109
XM_017029047.1 XP_016884536.1 4 - 5 932 109
XM_017029048.1 XP_016884537.1 4 - 5 932 109
XM_017029049.1 XP_016884538.1 4 - 5 837 109
XM_017029050.1 XP_016884539.1 4 - 5 932 109
XM_017029051.1 XP_016884540.1 4 - 5 837 109
XM_017029052.1 XP_016884541.1 4 - 5 932 109
XM_017029054.1 XP_016884543.1 4 - 5 932 109
XM_017029055.1 XP_016884544.1 3 - 4 841 109
XM_017029056.1 XP_016884545.1 4 - 5 932 109
XM_017029057.1 XP_016884546.1 4 - 5 932 109
XM_017029058.1 XP_016884547.1 4 - 5 815 109
XM_017029059.1 XP_016884548.1 4 - 5 841 109
XM_017029060.1 XP_016884549.1 4 - 5 840 109
XM_017029061.1 XP_016884550.1 4 - 5 932 109
XM_017029062.1 XP_016884551.1 3 - 4 841 109
XM_017029063.1 XP_016884552.1 4 - 5 841 109
XM_017029064.1 XP_016884553.1 3 - 4 724 109
GenBank mRNA AB051454.1 - 4 - 5 844 109
AK054717.1 - 2 - 3 365 109
AK057648.1 - 4 - 5 770 109
AK290211.1 - 4 - 5 861 109
AL365512.1 - 2 - 3 462 109
AL713795.1 - 4 - 5 890 109
AY043416.1 - 3 - 4 461 109
BC028625.1 - 4 - 5 846 109
BC050285.1 - 4 - 5 823 109
BC065030.1 - 3 - 4 663 109
CR456392.1 - 3 - 4 362 109

View Full Product Details