The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173354.3 | NP_775490.2 | 11 - 12 | 1589 | 73 |
XM_011529474.2 | XP_011527776.1 | 10 - 11 | 1446 | 73 | |
GenBank mRNA | AB047786.1 | - | 11 - 12 | 1589 | 73 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320643.1 | NP_001307572.1 | 11 - 12 | 1595 | 73 |
XM_017028208.1 | XP_016883697.1 | 10 - 11 | 1447 | 73 | |
GenBank mRNA | AK131076.1 | - | 10 - 11 | 1406 | 73 |
BC038504.1 | - | 11 - 12 | 1595 | 73 |