Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72228447-72228686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4881222 c.390T>A p.D130E Chr.11: 72228491-72228491 on Build GRCh38
7282713 c.378G>A p.P126P Chr.11: 72228479-72228479 on Build GRCh38
4882771 c.376C>A p.P126T Chr.11: 72228477-72228477 on Build GRCh38
1356787 c.361C>T p.R121* Chr.11: 72228462-72228462 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768475256 Chr.11: 72228505-72228505 on Build GRCh38
rs759655328 Chr.11: 72228532-72228532 on Build GRCh38
rs538502034 Chr.11: 72228548-72228548 on Build GRCh38
rs536046056 Chr.11: 72228617-72228617 on Build GRCh38
rs749225658 Chr.11: 72228495-72228495 on Build GRCh38
rs1048595584 Chr.11: 72228641-72228641 on Build GRCh38
rs748448649 Chr.11: 72228567-72228567 on Build GRCh38
rs568856228 Chr.11: 72228579-72228579 on Build GRCh38
rs918846796 Chr.11: 72228601-72228601 on Build GRCh38
rs776894351 Chr.11: 72228523-72228523 on Build GRCh38
rs930093088 Chr.11: 72228618-72228618 on Build GRCh38
rs202044589 Chr.11: 72228484-72228484 on Build GRCh38
rs200615694 Chr.11: 72228536-72228536 on Build GRCh38
rs764732670 Chr.11: 72228547-72228547 on Build GRCh38
rs371726889 Chr.11: 72228513-72228513 on Build GRCh38
rs756549919 Chr.11: 72228541-72228541 on Build GRCh38
rs920419704 Chr.11: 72228594-72228594 on Build GRCh38
rs1000207551 Chr.11: 72228509-72228509 on Build GRCh38
rs916614738 Chr.11: 72228552-72228552 on Build GRCh38
rs753208760 Chr.11: 72228468-72228468 on Build GRCh38
rs966495240 Chr.11: 72228531-72228531 on Build GRCh38
rs528612645 Chr.11: 72228478-72228478 on Build GRCh38
rs17847221 Chr.11: 72228551-72228551 on Build GRCh38
rs774083889 Chr.11: 72228507-72228507 on Build GRCh38
rs763316860 Chr.11: 72228546-72228546 on Build GRCh38
rs145795148 Chr.11: 72228479-72228479 on Build GRCh38
rs764410685 Chr.11: 72228475-72228475 on Build GRCh38
rs771083237 Chr.11: 72228521-72228521 on Build GRCh38
rs916336133 Chr.11: 72228525-72228525 on Build GRCh38
rs949650901 Chr.11: 72228560-72228560 on Build GRCh38
rs1048473891 Chr.11: 72228644-72228644 on Build GRCh38
rs139756712 Chr.11: 72228469-72228469 on Build GRCh38
rs745317721 Chr.11: 72228486-72228486 on Build GRCh38
rs765269255 Chr.11: 72228534-72228534 on Build GRCh38
rs1031114646 Chr.11: 72228510-72228512 on Build GRCh38
rs755641145 Chr.11: 72228487-72228487 on Build GRCh38

View Full Product Details