Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37861935-37862196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242147 c.281T>A p.I94N Chr.7: 37862038-37862038 on Build GRCh38
1089438 c.342C>T p.I114I Chr.7: 37862099-37862099 on Build GRCh38
421460 c.345T>C p.D115D Chr.7: 37862102-37862102 on Build GRCh38
6563938 c.311C>T p.P104L Chr.7: 37862068-37862068 on Build GRCh38
286215 c.312G>A p.P104P Chr.7: 37862069-37862069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191137504 Chr.7: 37862025-37862025 on Build GRCh38
rs1004372553 Chr.7: 37862040-37862040 on Build GRCh38
rs755343739 Chr.7: 37862166-37862166 on Build GRCh38
rs376045586 Chr.7: 37861977-37861977 on Build GRCh38
rs370705490 Chr.7: 37862013-37862013 on Build GRCh38
rs1016173782 Chr.7: 37862077-37862077 on Build GRCh38
rs201295247 Chr.7: 37862042-37862042 on Build GRCh38
rs968749440 Chr.7: 37862122-37862122 on Build GRCh38
rs752059115 Chr.7: 37861981-37861982 on Build GRCh38
rs910173023 Chr.7: 37861998-37861998 on Build GRCh38
rs151218163 Chr.7: 37862137-37862137 on Build GRCh38
rs749602472 Chr.7: 37862071-37862071 on Build GRCh38
rs760590328 Chr.7: 37862106-37862106 on Build GRCh38
rs1054608750 Chr.7: 37862003-37862003 on Build GRCh38
rs117149381 Chr.7: 37862001-37862001 on Build GRCh38
rs773106974 Chr.7: 37862016-37862016 on Build GRCh38
rs760643364 Chr.7: 37862017-37862017 on Build GRCh38
rs143447596 Chr.7: 37862100-37862100 on Build GRCh38
rs199734697 Chr.7: 37862099-37862099 on Build GRCh38
rs756726204 Chr.7: 37862155-37862155 on Build GRCh38
rs751058129 Chr.7: 37862151-37862151 on Build GRCh38
rs768937346 Chr.7: 37862084-37862084 on Build GRCh38
rs374907632 Chr.7: 37862136-37862136 on Build GRCh38
rs770842803 Chr.7: 37862124-37862124 on Build GRCh38
rs763685517 Chr.7: 37862149-37862149 on Build GRCh38
rs1052341809 Chr.7: 37861968-37861968 on Build GRCh38
rs536359204 Chr.7: 37862069-37862069 on Build GRCh38
rs376090348 Chr.7: 37862005-37862005 on Build GRCh38
rs2249451 Chr.7: 37861979-37861979 on Build GRCh38
rs567620217 Chr.7: 37862068-37862068 on Build GRCh38
rs368506712 Chr.7: 37862024-37862024 on Build GRCh38
rs762762070 Chr.7: 37862143-37862143 on Build GRCh38
rs868857499 Chr.7: 37862103-37862103 on Build GRCh38
rs757367181 Chr.7: 37862059-37862059 on Build GRCh38
rs781738300 Chr.7: 37862098-37862098 on Build GRCh38
rs772166327 Chr.7: 37862015-37862015 on Build GRCh38
rs372631973 Chr.7: 37862101-37862101 on Build GRCh38
rs887276229 Chr.7: 37862033-37862033 on Build GRCh38
rs368696812 Chr.7: 37862043-37862043 on Build GRCh38
rs778404063 Chr.7: 37861978-37861979 on Build GRCh38
rs145985042 Chr.7: 37862026-37862026 on Build GRCh38
rs759334409 Chr.7: 37862132-37862132 on Build GRCh38
rs180803661 Chr.7: 37862165-37862165 on Build GRCh38
rs776436705 Chr.7: 37862125-37862125 on Build GRCh38
rs376999057 Chr.7: 37862021-37862021 on Build GRCh38
rs139983071 Chr.7: 37862032-37862032 on Build GRCh38
rs780574300 Chr.7: 37862158-37862158 on Build GRCh38
rs770252954 Chr.7: 37861991-37861991 on Build GRCh38
rs201581622 Chr.7: 37862019-37862019 on Build GRCh38
rs75531720 Chr.7: 37862014-37862014 on Build GRCh38
rs781227597 Chr.7: 37862065-37862065 on Build GRCh38
rs759261323 Chr.7: 37862022-37862022 on Build GRCh38

View Full Product Details