Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27558288-27558561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6517025 c.805G>T p.E269* Chr.9: 27558541-27558541 on Build GRCh38
380700 c.796T>G p.C266G Chr.9: 27558550-27558550 on Build GRCh38
753687 c.850C>T p.L284L Chr.9: 27558496-27558496 on Build GRCh38
753688 c.851T>A p.L284Q Chr.9: 27558495-27558495 on Build GRCh38
6473117 c.822T>C p.Y274Y Chr.9: 27558524-27558524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907424383 Chr.9: 27558332-27558332 on Build GRCh38
rs747230783 Chr.9: 27558470-27558470 on Build GRCh38
rs762776605 Chr.9: 27558524-27558524 on Build GRCh38
rs371913416 Chr.9: 27558447-27558447 on Build GRCh38
rs911595933 Chr.9: 27558498-27558498 on Build GRCh38
rs764000894 Chr.9: 27558527-27558527 on Build GRCh38
rs374953135 Chr.9: 27558336-27558336 on Build GRCh38
rs960100300 Chr.9: 27558380-27558380 on Build GRCh38
rs1054714246 Chr.9: 27558381-27558381 on Build GRCh38
rs746018934 Chr.9: 27558486-27558486 on Build GRCh38
rs986186999 Chr.9: 27558481-27558481 on Build GRCh38
rs749328517 Chr.9: 27558346-27558346 on Build GRCh38
rs1001957237 Chr.9: 27558341-27558341 on Build GRCh38
rs771227536 Chr.9: 27558473-27558473 on Build GRCh38
rs961876718 Chr.9: 27558407-27558407 on Build GRCh38
rs560031223 Chr.9: 27558408-27558408 on Build GRCh38
rs769851288 Chr.9: 27558507-27558507 on Build GRCh38
rs775575174 Chr.9: 27558522-27558522 on Build GRCh38
rs776552699 Chr.9: 27558474-27558474 on Build GRCh38
rs992948437 Chr.9: 27558398-27558398 on Build GRCh38
rs1001763698 Chr.9: 27558345-27558345 on Build GRCh38
rs375808867 Chr.9: 27558461-27558461 on Build GRCh38
rs774015166 Chr.9: 27558533-27558533 on Build GRCh38
rs17696570 Chr.9: 27558439-27558439 on Build GRCh38
rs202082885 Chr.9: 27558402-27558402 on Build GRCh38
rs748505438 Chr.9: 27558452-27558452 on Build GRCh38
rs777924717 Chr.9: 27558462-27558462 on Build GRCh38
rs545433336 Chr.9: 27558537-27558537 on Build GRCh38

View Full Product Details