The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167576.1 | NP_001161048.1 | 8 - 9 | 2215 | 110 |
NM_001167577.1 | NP_001161049.1 | 9 - 10 | 2380 | 110 | |
NM_020389.2 | NP_065122.1 | 10 - 11 | 2563 | 110 | |
GenBank mRNA | AJ272034.1 | - | 9 - 10 | 2341 | 110 |
AJ421783.1 | - | 9 - 10 | 2341 | 110 | |
AJ549088.1 | - | 8 - 9 | 2158 | 110 | |
AJ549089.1 | - | 8 - 9 | 2176 | 110 | |
AJ549090.1 | - | 6 - 7 | 1776 | 110 | |
AY167926.1 | - | 8 - 9 | 2173 | 110 | |
AY167927.1 | - | 7 - 8 | 1993 | 110 | |
BC128185.1 | - | 9 - 10 | 2341 | 110 | |
BC128186.1 | - | 9 - 10 | 2357 | 110 | |
BI826927.1 | - | 3 - 4 | 340 | 110 |