The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001509.2 | NP_001500.1 | 1 - 2 | 95 | 74 |
NM_003996.3 | NP_003987.2 | 1 - 2 | 95 | 74 | |
GenBank mRNA | AJ005277.1 | - | 1 - 2 | 95 | 74 |
BC128159.1 | - | 1 - 2 | 95 | 74 | |
BC128160.1 | - | 1 - 2 | 95 | 74 | |
FJ460514.1 | - | 1 - 2 | 95 | 74 |