The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006556.3 | NP_006547.1 | 1 - 2 | 413 | 75 |
GenBank mRNA | AK312803.1 | - | 1 - 2 | 372 | 75 |
BC006089.1 | - | 1 - 2 | 396 | 75 | |
BC007694.2 | - | 1 - 2 | 326 | 75 | |
BQ061044.1 | - | 1 - 2 | 413 | 75 | |
BT019976.1 | - | 1 - 2 | 90 | 75 | |
CR541998.1 | - | 1 - 2 | 90 | 75 | |
L77213.1 | - | 1 - 2 | 125 | 75 |