The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007282.4 | NP_009213.1 | 2 - 3 | 765 | 66 |
XM_017005656.1 | XP_016861145.1 | 2 - 3 | 714 | 66 | |
XM_017005657.1 | XP_016861146.1 | 2 - 3 | 847 | 66 | |
GenBank mRNA | BC009803.1 | - | 2 - 3 | 243 | 66 |