Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102950031-102950304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5360774 c.875C>T p.A292V Chr.11: 102950152-102950152 on Build GRCh38
5768616 c.843A>G p.P281P Chr.11: 102950184-102950184 on Build GRCh38
2108578 c.867C>G p.S289S Chr.11: 102950160-102950160 on Build GRCh38
147315 c.800-9C>T p.? Chr.11: 102950236-102950236 on Build GRCh38
6066985 c.850T>C p.C284R Chr.11: 102950177-102950177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782608649 Chr.11: 102950243-102950243 on Build GRCh38
rs139253521 Chr.11: 102950076-102950076 on Build GRCh38
rs782776376 Chr.11: 102950078-102950078 on Build GRCh38
rs782702393 Chr.11: 102950196-102950196 on Build GRCh38
rs782705544 Chr.11: 102950075-102950075 on Build GRCh38
rs376033599 Chr.11: 102950223-102950223 on Build GRCh38
rs369619125 Chr.11: 102950081-102950081 on Build GRCh38
rs782338553 Chr.11: 102950118-102950118 on Build GRCh38
rs148230434 Chr.11: 102950185-102950185 on Build GRCh38
rs199680440 Chr.11: 102950112-102950112 on Build GRCh38
rs782210300 Chr.11: 102950122-102950122 on Build GRCh38
rs201029263 Chr.11: 102950234-102950234 on Build GRCh38
rs782433681 Chr.11: 102950105-102950105 on Build GRCh38
rs782228973 Chr.11: 102950069-102950070 on Build GRCh38
rs781841656 Chr.11: 102950209-102950209 on Build GRCh38
rs979816589 Chr.11: 102950126-102950126 on Build GRCh38
rs782353607 Chr.11: 102950181-102950181 on Build GRCh38
rs782684582 Chr.11: 102950254-102950254 on Build GRCh38
rs782239802 Chr.11: 102950177-102950177 on Build GRCh38
rs367735577 Chr.11: 102950255-102950255 on Build GRCh38
rs782547926 Chr.11: 102950091-102950091 on Build GRCh38
rs782777750 Chr.11: 102950220-102950220 on Build GRCh38
rs781808044 Chr.11: 102950099-102950099 on Build GRCh38
rs782527417 Chr.11: 102950137-102950137 on Build GRCh38
rs991340696 Chr.11: 102950070-102950070 on Build GRCh38
rs782269102 Chr.11: 102950245-102950245 on Build GRCh38
rs782394851 Chr.11: 102950160-102950160 on Build GRCh38
rs782018679 Chr.11: 102950188-102950188 on Build GRCh38
rs782620150 Chr.11: 102950143-102950143 on Build GRCh38
rs370275782 Chr.11: 102950235-102950235 on Build GRCh38
rs149330684 Chr.11: 102950171-102950171 on Build GRCh38
rs782322194 Chr.11: 102950259-102950259 on Build GRCh38
rs17860556 Chr.11: 102950236-102950236 on Build GRCh38
rs781812031 Chr.11: 102950068-102950068 on Build GRCh38
rs782083671 Chr.11: 102950210-102950210 on Build GRCh38
rs782380897 Chr.11: 102950248-102950248 on Build GRCh38
rs376304010 Chr.11: 102950077-102950077 on Build GRCh38
rs370869245 Chr.11: 102950193-102950193 on Build GRCh38
rs782286302 Chr.11: 102950153-102950153 on Build GRCh38

View Full Product Details