Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72228630-72228883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6480051 c.414G>A p.K138K Chr.11: 72228743-72228743 on Build GRCh38
5601636 c.456C>T p.A152A Chr.11: 72228785-72228785 on Build GRCh38
3452986 c.418C>T p.P140S Chr.11: 72228747-72228747 on Build GRCh38
931660 c.491A>C p.E164A Chr.11: 72228820-72228820 on Build GRCh38
195545 c.429G>A p.P143P Chr.11: 72228758-72228758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755620537 Chr.11: 72228700-72228700 on Build GRCh38
rs549746015 Chr.11: 72228674-72228674 on Build GRCh38
rs146603018 Chr.11: 72228737-72228737 on Build GRCh38
rs778377922 Chr.11: 72228853-72228854 on Build GRCh38
rs199991260 Chr.11: 72228833-72228833 on Build GRCh38
rs768035194 Chr.11: 72228780-72228780 on Build GRCh38
rs201835641 Chr.11: 72228856-72228856 on Build GRCh38
rs770179572 Chr.11: 72228715-72228715 on Build GRCh38
rs749317647 Chr.11: 72228855-72228855 on Build GRCh38
rs999772869 Chr.11: 72228829-72228829 on Build GRCh38
rs778834790 Chr.11: 72228749-72228749 on Build GRCh38
rs752670331 Chr.11: 72228811-72228811 on Build GRCh38
rs758164532 Chr.11: 72228816-72228816 on Build GRCh38
rs753229253 Chr.11: 72228798-72228798 on Build GRCh38
rs902447558 Chr.11: 72228823-72228823 on Build GRCh38
rs61735633 Chr.11: 72228830-72228830 on Build GRCh38
rs766088509 Chr.11: 72228796-72228796 on Build GRCh38
rs760197203 Chr.11: 72228794-72228794 on Build GRCh38
rs1030220603 Chr.11: 72228793-72228793 on Build GRCh38
rs1041030521 Chr.11: 72228748-72228748 on Build GRCh38
rs113871062 Chr.11: 72228788-72228788 on Build GRCh38
rs762267711 Chr.11: 72228779-72228779 on Build GRCh38
rs773663316 Chr.11: 72228760-72228760 on Build GRCh38
rs779479923 Chr.11: 72228706-72228706 on Build GRCh38
rs368460863 Chr.11: 72228810-72228810 on Build GRCh38
rs56353029 Chr.11: 72228861-72228861 on Build GRCh38
rs767268219 Chr.11: 72228691-72228691 on Build GRCh38
rs370550151 Chr.11: 72228857-72228857 on Build GRCh38
rs778816094 Chr.11: 72228807-72228807 on Build GRCh38
rs376049368 Chr.11: 72228753-72228753 on Build GRCh38
rs771919378 Chr.11: 72228755-72228755 on Build GRCh38
rs1032175507 Chr.11: 72228854-72228854 on Build GRCh38
rs572825150 Chr.11: 72228773-72228773 on Build GRCh38
rs201929168 Chr.11: 72228834-72228834 on Build GRCh38
rs781357595 Chr.11: 72228757-72228757 on Build GRCh38
rs999276105 Chr.11: 72228825-72228825 on Build GRCh38
rs769232228 Chr.11: 72228768-72228768 on Build GRCh38
rs746849326 Chr.11: 72228828-72228828 on Build GRCh38
rs887520792 Chr.11: 72228667-72228667 on Build GRCh38
rs753411829 Chr.11: 72228729-72228729 on Build GRCh38
rs141059764 Chr.11: 72228765-72228765 on Build GRCh38
rs201045134 Chr.11: 72228772-72228772 on Build GRCh38
rs754540210 Chr.11: 72228802-72228802 on Build GRCh38
rs777492773 Chr.11: 72228827-72228827 on Build GRCh38
rs376366065 Chr.11: 72228696-72228696 on Build GRCh38
rs942176300 Chr.11: 72228850-72228850 on Build GRCh38

View Full Product Details