Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6012719-6012978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465788 c.808A>C p.R270R Chr.10: 6012883-6012883 on Build GRCh38
7285519 c.795G>A p.Q265Q Chr.10: 6012896-6012896 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769795224 Chr.10: 6012843-6012843 on Build GRCh38
rs886047080 Chr.10: 6012769-6012769 on Build GRCh38
rs569908109 Chr.10: 6012930-6012930 on Build GRCh38
rs753838977 Chr.10: 6012773-6012773 on Build GRCh38
rs747854116 Chr.10: 6012835-6012835 on Build GRCh38
rs750285734 Chr.10: 6012903-6012903 on Build GRCh38
rs12722714 Chr.10: 6012765-6012765 on Build GRCh38
rs775980513 Chr.10: 6012878-6012878 on Build GRCh38
rs747799216 Chr.10: 6012927-6012927 on Build GRCh38
rs547047548 Chr.10: 6012811-6012811 on Build GRCh38
rs972136274 Chr.10: 6012780-6012780 on Build GRCh38
rs12722600 Chr.10: 6012802-6012802 on Build GRCh38
rs372312640 Chr.10: 6012836-6012836 on Build GRCh38
rs758129175 Chr.10: 6012929-6012929 on Build GRCh38
rs749452297 Chr.10: 6012768-6012768 on Build GRCh38
rs778609114 Chr.10: 6012924-6012924 on Build GRCh38
rs935037648 Chr.10: 6012943-6012943 on Build GRCh38
rs886047081 Chr.10: 6012779-6012779 on Build GRCh38
rs767207934 Chr.10: 6012896-6012896 on Build GRCh38
rs920592834 Chr.10: 6012865-6012865 on Build GRCh38
rs762011738 Chr.10: 6012949-6012950 on Build GRCh38
rs12722712 Chr.10: 6012876-6012876 on Build GRCh38
rs746177077 Chr.10: 6012936-6012936 on Build GRCh38
rs752490211 Chr.10: 6012868-6012868 on Build GRCh38
rs774531130 Chr.10: 6012885-6012885 on Build GRCh38
rs12722713 Chr.10: 6012817-6012817 on Build GRCh38
rs762059830 Chr.10: 6012887-6012887 on Build GRCh38
rs575663406 Chr.10: 6012822-6012822 on Build GRCh38
rs12722599 Chr.10: 6012950-6012950 on Build GRCh38
rs757598971 Chr.10: 6012889-6012891 on Build GRCh38
rs770086074 Chr.10: 6012937-6012937 on Build GRCh38
rs1012922758 Chr.10: 6012755-6012755 on Build GRCh38
rs549892288 Chr.10: 6012749-6012749 on Build GRCh38
rs962415501 Chr.10: 6012769-6012769 on Build GRCh38
rs781530309 Chr.10: 6012943-6012943 on Build GRCh38
rs970867521 Chr.10: 6012779-6012779 on Build GRCh38
rs762979594 Chr.10: 6012879-6012879 on Build GRCh38
rs778662335 Chr.10: 6012826-6012826 on Build GRCh38
rs548889439 Chr.10: 6012912-6012912 on Build GRCh38
rs914624268 Chr.10: 6012950-6012950 on Build GRCh38
rs369719952 Chr.10: 6012920-6012920 on Build GRCh38
rs74162101 Chr.10: 6012919-6012919 on Build GRCh38
rs746347754 Chr.10: 6012859-6012859 on Build GRCh38

View Full Product Details