Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50929384-50929657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559814 c.179T>A p.L60Q Chr.13: 50929517-50929517 on Build GRCh38
6569555 c.150T>C p.I50I Chr.13: 50929488-50929488 on Build GRCh38
6553966 c.177G>T p.Q59H Chr.13: 50929515-50929515 on Build GRCh38
5499169 c.138_140delAGA p.E47delE Chr.13: 50929476-50929478 on Build GRCh38
6767099 c.159C>A p.F53L Chr.13: 50929497-50929497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759876767 Chr.13: 50929624-50929624 on Build GRCh38
rs758911416 Chr.13: 50929531-50929531 on Build GRCh38
rs747145612 Chr.13: 50929627-50929628 on Build GRCh38
rs371461579 Chr.13: 50929585-50929585 on Build GRCh38
rs777020973 Chr.13: 50929614-50929614 on Build GRCh38
rs148811739 Chr.13: 50929541-50929541 on Build GRCh38
rs145465454 Chr.13: 50929540-50929540 on Build GRCh38
rs779094485 Chr.13: 50929430-50929430 on Build GRCh38
rs75325951 Chr.13: 50929517-50929517 on Build GRCh38
rs199744138 Chr.13: 50929424-50929425 on Build GRCh38
rs759251667 Chr.13: 50929511-50929511 on Build GRCh38
rs768983041 Chr.13: 50929441-50929441 on Build GRCh38
rs770080674 Chr.13: 50929499-50929499 on Build GRCh38
rs749691175 Chr.13: 50929451-50929451 on Build GRCh38
rs980587217 Chr.13: 50929415-50929415 on Build GRCh38
rs761230115 Chr.13: 50929609-50929609 on Build GRCh38
rs748268980 Chr.13: 50929591-50929591 on Build GRCh38
rs762449221 Chr.13: 50929633-50929633 on Build GRCh38
rs374465686 Chr.13: 50929502-50929502 on Build GRCh38
rs757200430 Chr.13: 50929572-50929572 on Build GRCh38
rs779173843 Chr.13: 50929586-50929586 on Build GRCh38
rs780704541 Chr.13: 50929580-50929581 on Build GRCh38
rs763195911 Chr.13: 50929425-50929426 on Build GRCh38
rs371087934 Chr.13: 50929428-50929428 on Build GRCh38
rs78847362 Chr.13: 50929583-50929583 on Build GRCh38
rs537203779 Chr.13: 50929508-50929508 on Build GRCh38
rs542252075 Chr.13: 50929612-50929612 on Build GRCh38
rs750984942 Chr.13: 50929455-50929458 on Build GRCh38
rs35416748 Chr.13: 50929494-50929494 on Build GRCh38
rs772503413 Chr.13: 50929491-50929491 on Build GRCh38
rs556877869 Chr.13: 50929518-50929518 on Build GRCh38
rs373866654 Chr.13: 50929438-50929439 on Build GRCh38
rs752428694 Chr.13: 50929631-50929631 on Build GRCh38
rs982668471 Chr.13: 50929432-50929432 on Build GRCh38
rs765474219 Chr.13: 50929630-50929630 on Build GRCh38
rs368589166 Chr.13: 50929521-50929521 on Build GRCh38
rs912717939 Chr.13: 50929431-50929431 on Build GRCh38
rs35595258 Chr.13: 50929527-50929527 on Build GRCh38
rs75326546 Chr.13: 50929510-50929510 on Build GRCh38
rs201814710 Chr.13: 50929456-50929456 on Build GRCh38
rs781187392 Chr.13: 50929578-50929578 on Build GRCh38
rs573238722 Chr.13: 50929603-50929603 on Build GRCh38
rs78071087 Chr.13: 50929556-50929556 on Build GRCh38
rs749531762 Chr.13: 50929436-50929436 on Build GRCh38
rs921607762 Chr.13: 50929498-50929498 on Build GRCh38
rs541903338 Chr.13: 50929531-50929531 on Build GRCh38
rs528197168 Chr.13: 50929411-50929412 on Build GRCh38
rs141881672 Chr.13: 50929515-50929515 on Build GRCh38
rs534340192 Chr.13: 50929430-50929430 on Build GRCh38
rs761921568 Chr.13: 50929465-50929465 on Build GRCh38
rs776551576 Chr.13: 50929426-50929427 on Build GRCh38
rs772222915 Chr.13: 50929594-50929594 on Build GRCh38

View Full Product Details