Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96184818-96185079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
UniGene
Hs.489190
Gene Chromosome Location
Chr.7: 96120220-96322147 on Build GRCh38
Transcripts
NM_001160210.1 NM_014251.2 NR_027662.1 XM_006715831.3 XM_011515727.2 XM_017011663.1 XM_017011664.1 XM_017011665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093388 c.983C>T p.S328L Chr.7: 96184962-96184962 on Build GRCh38
1093387 c.1011T>C p.V337V Chr.7: 96184934-96184934 on Build GRCh38
3883585 c.972A>G p.Q324Q Chr.7: 96184973-96184973 on Build GRCh38
1496705 c.977C>G p.A326G Chr.7: 96184968-96184968 on Build GRCh38
6780523 c.955C>T p.R319* Chr.7: 96184990-96184990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766917149 Chr.7: 96184999-96184999 on Build GRCh38
rs368612322 Chr.7: 96185027-96185027 on Build GRCh38
rs539240952 Chr.7: 96184905-96184905 on Build GRCh38
rs760796001 Chr.7: 96184888-96184888 on Build GRCh38
rs769297213 Chr.7: 96184945-96184945 on Build GRCh38
rs770931172 Chr.7: 96184947-96184947 on Build GRCh38
rs752114544 Chr.7: 96185003-96185003 on Build GRCh38
rs373818105 Chr.7: 96184924-96184924 on Build GRCh38
rs1042155275 Chr.7: 96184901-96184901 on Build GRCh38
rs777248648 Chr.7: 96185041-96185041 on Build GRCh38
rs1001025380 Chr.7: 96184957-96184957 on Build GRCh38
rs763191789 Chr.7: 96184990-96184990 on Build GRCh38
rs755812402 Chr.7: 96185037-96185037 on Build GRCh38
rs767051127 Chr.7: 96185014-96185014 on Build GRCh38
rs755469215 Chr.7: 96185007-96185007 on Build GRCh38
rs765358773 Chr.7: 96184985-96184985 on Build GRCh38
rs374017898 Chr.7: 96184894-96184894 on Build GRCh38
rs775662045 Chr.7: 96184883-96184883 on Build GRCh38
rs760851084 Chr.7: 96184961-96184961 on Build GRCh38
rs1013757741 Chr.7: 96185006-96185006 on Build GRCh38
rs111325365 Chr.7: 96184889-96184889 on Build GRCh38
rs113850932 Chr.7: 96184979-96184979 on Build GRCh38
rs754199361 Chr.7: 96184861-96184861 on Build GRCh38
rs748986342 Chr.7: 96184946-96184946 on Build GRCh38
rs762114824 Chr.7: 96184893-96184893 on Build GRCh38
rs751105436 Chr.7: 96184917-96184917 on Build GRCh38
rs747861059 Chr.7: 96184939-96184939 on Build GRCh38
rs780663014 Chr.7: 96184937-96184937 on Build GRCh38
rs534585904 Chr.7: 96184955-96184955 on Build GRCh38
rs771449632 Chr.7: 96184989-96184989 on Build GRCh38
rs895069432 Chr.7: 96184920-96184920 on Build GRCh38
rs56210033 Chr.7: 96184881-96184881 on Build GRCh38
rs762137400 Chr.7: 96184976-96184976 on Build GRCh38
rs1012084121 Chr.7: 96184975-96184975 on Build GRCh38
rs764401478 Chr.7: 96184964-96184965 on Build GRCh38
rs970558852 Chr.7: 96184991-96184991 on Build GRCh38
rs752177815 Chr.7: 96185019-96185019 on Build GRCh38
rs768915439 Chr.7: 96184964-96184964 on Build GRCh38
rs764417564 Chr.7: 96184892-96184892 on Build GRCh38
rs763288993 Chr.7: 96184893-96184894 on Build GRCh38
rs933074073 Chr.7: 96184858-96184858 on Build GRCh38
rs776848302 Chr.7: 96184967-96184967 on Build GRCh38
rs921124221 Chr.7: 96184852-96184852 on Build GRCh38
rs758996304 Chr.7: 96184903-96184903 on Build GRCh38
rs76371700 Chr.7: 96184896-96184896 on Build GRCh38

View Full Product Details