The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007129.3 | NP_009060.2 | 1 - 2 | 1364 | 72 |
XM_011521110.2 | XP_011519412.1 | 1 - 2 | 1135 | 72 | |
GenBank mRNA | AF104902.1 | - | 1 - 2 | 1074 | 72 |
AF193855.1 | - | 1 - 2 | 1071 | 72 |