The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204182.1 | NP_001191111.1 | 4 - 5 | 790 | 124 |
NM_001204183.1 | NP_001191112.1 | 4 - 5 | 790 | 124 | |
NM_002497.3 | NP_002488.1 | 4 - 5 | 790 | 124 | |
XM_005273147.1 | XP_005273204.1 | 4 - 5 | 787 | 124 | |
GenBank mRNA | AK223353.1 | - | 4 - 5 | 751 | 124 |
AK225522.1 | - | 4 - 5 | 751 | 124 | |
AY045701.1 | - | 4 - 5 | 721 | 124 | |
BC043502.1 | - | 4 - 5 | 778 | 124 | |
BC052807.1 | - | 4 - 5 | 785 | 124 | |
BT019729.1 | - | 4 - 5 | 639 | 124 | |
U11050.1 | - | 4 - 5 | 773 | 124 | |
Z29066.1 | - | 4 - 5 | 721 | 124 |