The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014591.4 | NP_055406.2 | 9 - 10 | 1164 | 146 |
NM_173191.2 | NP_775283.1 | 9 - 10 | 1119 | 146 | |
NM_173192.2 | NP_775284.1 | 8 - 9 | 1065 | 146 | |
NM_173193.2 | NP_775285.1 | 7 - 8 | 990 | 146 | |
NM_173194.2 | NP_775286.1 | 7 - 8 | 646 | 146 | |
NM_173195.2 | NP_775287.1 | 7 - 8 | 969 | 146 | |
XM_017016161.1 | XP_016871650.1 | 7 - 8 | 716 | 146 | |
GenBank mRNA | AJ276317.1 | - | 2 - 3 | 132 | 145 |
AK027347.1 | - | 7 - 8 | 637 | 146 | |
AK055089.1 | - | 9 - 10 | 1043 | 146 | |
AK094668.1 | - | 11 - 12 | 1171 | 146 | |
AK125382.1 | - | 7 - 8 | 1676 | 146 | |
AK295324.1 | - | 5 - 6 | 600 | 146 | |
AK309368.1 | - | 7 - 8 | 845 | 146 | |
BC034685.1 | - | 8 - 9 | 928 | 146 |