The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256135.1 | NP_001243064.1 | 4 - 5 | 511 | 93 |
NM_001316.3 | NP_001307.2 | 4 - 5 | 511 | 93 | |
XM_011528599.2 | XP_011526901.1 | 4 - 5 | 445 | 93 | |
GenBank mRNA | AF053640.1 | - | 4 - 5 | 418 | 93 |
AF053641.1 | - | 4 - 5 | 445 | 93 | |
AK297270.1 | - | 4 - 5 | 439 | 93 | |
AK297314.1 | - | 3 - 4 | 344 | 93 | |
AK312306.1 | - | 4 - 5 | 442 | 93 | |
BC108309.1 | - | 4 - 5 | 457 | 93 | |
BC109313.2 | - | 4 - 5 | 424 | 93 | |
BC109314.2 | - | 4 - 5 | 424 | 93 | |
BG392946.1 | - | 4 - 5 | 455 | 94 | |
DC402014.1 | - | 4 - 5 | 511 | 93 | |
EF426455.1 | - | 3 - 4 | 322 | 93 | |
U33286.1 | - | 4 - 5 | 445 | 93 |