Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10A5
Entrez Gene ID
144124
Gene Name
olfactory receptor family 10 subfamily A member 5
Gene Aliases
JCG6, OR10A1, OR11-403
Chromosome Location
Chr.11: 6845683 - 6846636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178168.1 NP_835462.1 1 - 1 244 65
GenBank mRNA AF324499.1 - 1 - 1 244 65
BC093651.1 - 1 - 1 275 65
BC093653.1 - 1 - 1 275 65

Gene Symbol
OR10A2
Entrez Gene ID
341276
Gene Name
olfactory receptor family 10 subfamily A member 2
Gene Aliases
OR10A2P, OR11-82, OR11-86, OST363
Chromosome Location
Chr.11: 6869755 - 6870666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004460.1 NP_001004460.1 1 - 1 202 65
GenBank mRNA BC136955.1 - 1 - 1 264 65

View Full Product Details