Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23917813-23918064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL15
Entrez Gene ID
6138
Gene Name
ribosomal protein L15
Gene Aliases
DBA12, EC45, L15, RPL10, RPLY10, RPYL10
UniGene
Hs.381219
Gene Chromosome Location
Chr.3: 23916804-23923696 on Build GRCh38
Transcripts
NM_001253379.1 NM_001253380.1 NM_001253382.1 NM_001253383.1 NM_001253384.1 NM_002948.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150982 c.131G>A p.R44Q Chr.3: 23917990-23917990 on Build GRCh38
7388130 c.72G>T p.R24S Chr.3: 23917931-23917931 on Build GRCh38
1309007 c.23A>G p.Q8R Chr.3: 23917882-23917882 on Build GRCh38
7315191 c.62T>A p.F21Y Chr.3: 23917921-23917921 on Build GRCh38
6280427 c.36A>G p.R12R Chr.3: 23917895-23917895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749731223 Chr.3: 23918015-23918015 on Build GRCh38
rs756448980 Chr.3: 23917855-23917855 on Build GRCh38
rs571705839 Chr.3: 23917982-23917982 on Build GRCh38
rs774562896 Chr.3: 23917877-23917877 on Build GRCh38
rs372215459 Chr.3: 23917852-23917852 on Build GRCh38
rs771618191 Chr.3: 23917870-23917870 on Build GRCh38
rs35629664 Chr.3: 23917934-23917934 on Build GRCh38
rs751126186 Chr.3: 23917845-23917845 on Build GRCh38
rs199591542 Chr.3: 23917846-23917846 on Build GRCh38
rs766954372 Chr.3: 23917987-23917987 on Build GRCh38
rs772286584 Chr.3: 23917914-23917914 on Build GRCh38
rs746395128 Chr.3: 23918041-23918041 on Build GRCh38
rs757838005 Chr.3: 23918017-23918017 on Build GRCh38
rs935839158 Chr.3: 23917904-23917904 on Build GRCh38
rs772231304 Chr.3: 23918043-23918043 on Build GRCh38
rs778025718 Chr.3: 23917856-23917856 on Build GRCh38
rs762863163 Chr.3: 23917940-23917940 on Build GRCh38
rs751502211 Chr.3: 23917979-23917979 on Build GRCh38
rs752350791 Chr.3: 23917988-23917988 on Build GRCh38
rs755759871 Chr.3: 23917991-23917991 on Build GRCh38
rs766367909 Chr.3: 23917960-23917960 on Build GRCh38
rs369956511 Chr.3: 23918036-23918036 on Build GRCh38
rs775725406 Chr.3: 23917925-23917925 on Build GRCh38
rs778175191 Chr.3: 23917997-23917997 on Build GRCh38
rs746016083 Chr.3: 23917906-23917906 on Build GRCh38
rs567287618 Chr.3: 23917859-23917859 on Build GRCh38
rs747767933 Chr.3: 23917854-23917854 on Build GRCh38
rs537969564 Chr.3: 23917880-23917880 on Build GRCh38
rs772921805 Chr.3: 23917937-23917937 on Build GRCh38

View Full Product Details