The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018117.11 | NP_060587.8 | 22 - 23 | 3000 | 82 |
XM_005269962.2 | XP_005270019.1 | 22 - 23 | 2717 | 82 | |
XM_005269963.2 | XP_005270020.1 | 22 - 23 | 2849 | 82 | |
XM_005269964.2 | XP_005270021.1 | 15 - 16 | 1755 | 82 | |
XM_017016397.1 | XP_016871886.1 | 21 - 22 | 2688 | 82 | |
XM_017016398.1 | XP_016871887.1 | 21 - 22 | 2618 | 82 | |
XM_017016399.1 | XP_016871888.1 | 21 - 22 | 2562 | 82 | |
GenBank mRNA | AB037772.2 | - | 22 - 23 | 2813 | 82 |
AF320223.1 | - | 22 - 23 | 2787 | 82 | |
AK000579.1 | - | 8 - 9 | 802 | 82 | |
AK001368.1 | - | 17 - 18 | 1903 | 82 | |
AK025050.1 | - | 5 - 6 | 511 | 82 | |
AK124522.1 | - | 9 - 10 | 3187 | 82 | |
AK299524.1 | - | 15 - 16 | 1620 | 82 | |
AK315145.1 | - | 22 - 23 | 2778 | 82 | |
AL137699.1 | - | 20 - 21 | 2473 | 82 | |
BC040469.1 | - | 22 - 23 | 2817 | 82 | |
BC071564.1 | - | 22 - 23 | 2783 | 82 |