Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr.10: 120851175 - 120909526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018117.11 NP_060587.8 2 - 3 445 100
XM_005269963.2 XP_005270020.1 2 - 3 294 100
XM_017016397.1 XP_016871886.1 2 - 3 307 100
XM_017016398.1 XP_016871887.1 2 - 3 237 100
XM_017016399.1 XP_016871888.1 2 - 3 237 100
XM_017016400.1 XP_016871889.1 2 - 3 336 100
GenBank mRNA AB037772.2 - 2 - 3 258 100
AF320223.1 - 2 - 3 232 100
AK301761.1 - 2 - 3 228 100
AK315145.1 - 2 - 3 223 100
BC040469.1 - 2 - 3 262 100
BC071564.1 - 2 - 3 228 100
BI912281.1 - 2 - 3 465 100
DC342128.1 - 2 - 3 445 100

View Full Product Details