The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000056.4 | NP_000047.1 | 5 - 6 | 713 | 83 |
NM_001318975.1 | NP_001305904.1 | 5 - 6 | 507 | 83 | |
NM_183050.3 | NP_898871.1 | 5 - 6 | 713 | 83 | |
XM_005248756.4 | XP_005248813.1 | 5 - 6 | 662 | 83 | |
XM_011536023.2 | XP_011534325.1 | 5 - 6 | 662 | 83 | |
XM_011536024.2 | XP_011534326.1 | 5 - 6 | 662 | 83 | |
XM_011536025.2 | XP_011534327.1 | 5 - 6 | 662 | 83 | |
XM_011536026.2 | XP_011534328.1 | 5 - 6 | 649 | 83 | |
GenBank mRNA | AK289977.1 | - | 5 - 6 | 642 | 83 |
AK304353.1 | - | 5 - 6 | 478 | 83 | |
AK310838.1 | - | 3 - 4 | 471 | 83 | |
AK316545.1 | - | 6 - 7 | 626 | 83 | |
BM553932.1 | - | 1 - 2 | 70 | 83 | |
BT020063.1 | - | 5 - 6 | 629 | 83 | |
M55575.1 | - | 5 - 6 | 633 | 83 | |
U50708.1 | - | 5 - 6 | 676 | 83 | |
X52446.1 | - | 5 - 6 | 572 | 83 |